Create MyQubit .qbt files for the Qubit 3 and 4 Fluorometer
This web tool will help you to create custom MyQubit assay .qbt files that you can upload to your Qubit 3 and 4 Fluorometer. If the assay you're creating does not require calibration, simply enter the name your assay then select the appropriate excitation and emission parameters. For assays that require calibration, we recommend using the “MyQubit EZ Guide” Excel® spreadsheet to help you generate the necessary constants. For complete instructions on how to use the MyQubit EZ Guide to generate constants and how to upload .qbt files to your Qubit 3 and 4 Fluorometer, see MyQubit Custom Assays on the Qubit 3 and 4 Fluorometer.
Please note that this web tool creates .qbt files that can only be used on the Qubit 3 and 4 Fluorometer.
To create .qbt files for the Qubit 2.0 Fluorometer, see MyQubit Custom Assays on the Qubit 2.0 Fluorometer for detailed instructions.
We hope to have this resolved soon.
For Research Use Only. Not for use in diagnostic procedures.