Rapid and accurate metal analysis
Enhance your lab efficiency and productivity with comprehensive software solutions for the complete analysis of a wide range of materials. Whether you have an optical emission spectrometry (OES) system or an X-ray fluorescence (XRF) system, we offer solutions to perform high-quality analysis easily and quickly.
OXSAS software versions for XRF and OES
Thermo Scientific OXSAS software for XRF instruments performs rapid, high-quality sample analysis using template-driven operations. Optional standard-less quantitative analysis packages offer additional versatility for all laboratories that analyze a wide range of materials. XRF analysis assures high repeatability which makes the technique effective for the control of major elements and expensive alloying elements.
Thermo Scientific OXSAS software for OES instruments runs a variety of analyses from simple tasks to complex jobs, including the development of new analytical methods and calibrations as well as rapid high-quality routine elemental analysis. OES provides rapid analysis of carbon and low nitrogen along with all other alloying elements in iron and steel while XRF offers most reliable and repeatable analysis of metallurgical slags, raw materials, including iron ores and sinters and cast irons.
OXSAS software is compatible with these Thermo Scientific analyzers:
The Thermo Scientific ARL OPTIM'X WDXRF Spectrometer analyzes elements from carbon to uranium in a variety of samples with accuracy, precision and reliability.
The Thermo Scientific ARL QUANT'X EDXRF Spectrometer provides major, minor and trace element quantification across a broad range of samples in minutes.
Solve analytical challenges in a wide range of applications with one comprehensive tool - the Thermo Scientific ARL PERFORM'X Sequential X-Ray Fluorescence Spectrometer for advanced materials characterization.
The Thermo Scientific ARL SMS-3500 Automation for Twin OES or XRF simplifies sample handling by providing a single introduction location for all samples with a magazine storage function to free the introduction.
The Thermo Scientific ARL SMS-2500 Robotics-based Automation for OES is an automated quality control solution to optimize your steel production operation.
The Thermo Scientific ARL SMS-2500 Robotics-based Automation for XRF is an automated solution for XRF elemental analysis of iron and steel.
The Thermo Scientific ARL Optical Emission Spectrometer provides the solution for routine elemental analysis, metals research and development.
The Thermo Scientific ARL 9900 Series combines fixed channels and one or two goniometers to ensure rapid and simultaneous analysis.