Search Thermo Fisher Scientific
NOTE: This map applies only to our new and improved pSilencer 2.1-U6 neo vector which began shipping in Dec 2003 (Lots 113P06 & 113P07).
This map and the annotated sequence below portrays the circular negative control vector provided with the kit. It includes the negative control siRNA template (66 base pairs) between the BamH I and Hind III sites.
We recommend the following primers for sequencing:
M13F(-40): 5'-GTTTTCCCAGTCACGAC-3'
Rev: 5'-GAGTTAGCTCACTCATTAGGC-3'
Site | Enzyme |
---|---|
4456 | AatII |
1151 | AflIII |
2044 | AhdI |
1567 | AlwNI |
4008 | AvaI |
4029 | AvrII |
730 | BamHI |
3462 | BanII |
2549 | BcgI |
2583 | BcgI' |
3964 | BsaBI |
2105 | BsaI |
1026 | BsaXI |
996 | BsaXI` |
4039 | BseRI |
45 | BsmBI |
2903 | BsmI |
436 | DraIII |
3913 | EagI |
396 | EcoRI |
792 | HindIII |
3741 | MscI |
3139 | NruI |
1151 | PciI |
436 | PmlI |
2962 | PsiI |
3744 | PstI |
3304 | RsrII |
2524 | ScaI |
4261 | SexAI |
4010 | SmaI |
4034 | StuI |
3704 | Tth111I |
4008 | XmaI |
2643 | XmnI |
4454 | ZraI |
Enzyme |
---|
AarI |
Acc65I |
AccI |
AfeI |
AflII |
AgeI |
AleI |
AloI |
AloI' |
ApaI |
AscI |
AsiSI |
BaeI |
BaeI' |
BbsI |
BbvCI |
BglII |
BlpI |
BmgBI |
BmtI |
Bpu10I |
BsgI |
BsiWI |
BspEI |
BsrGI |
BstBI |
BstEII |
BstXI |
BstZ17I |
Bsu36I |
BtrI |
ClaI |
EcoICRI |
EcoNI |
EcoRV |
FseI |
FspAI |
HincII |
HpaI |
KpnI |
MfeI |
MluI |
NheI |
NotI |
PacI |
PflMI |
PmeI |
PpuMI |
PshAI |
PspOMI |
PsrI |
PsrI' |
SacI |
SacII |
SalI |
SanDI |
SbfI |
SfiI |
SgfI |
SgrAI |
SnaBI |
SpeI |
SrfI |
SwaI |
TaqII' |
XbaI |
XcmI |
XhoI |