Search Thermo Fisher Scientific
Our long list of innovative products translates into multiple benefits for your team, enhancing effectiveness, efficiency and productivity.
Our shakers' triple eccentric drive mechanism provides 24-hour operation and allows you to shake large and uneven loads smoothly and uniformly. Plus, it’s guaranteed for life. Bench-top, floor, and stackable models are available, which are fully equipped with incubation, refrigeration, and water bath options to accommodate many applications and budgets.