With temperature security, quick recovery, and reliable validation, Thermo Scientific Ultra-Low Temperature (ULT) freezers are designed to deliver confidence — all while reducing your energy footprint. And, with a large team of field based service engineers and tech support specialists, we are here to assist should the need arise.
Learn more about your coverage
Field Service, Technical Support, and Depot Repair
Our team of experts is here to support you should any issue arise with your Thermo Scientific ULT. Our wide range of service solutions enables minimal downtime, allowing you to the peace of mind that your samples are safe.
Order service parts for your ULT
Visit our parts and accessories site to order any spare parts needed for your unit.
Care and Feeding: Tips and Tricks
ULT Freezers require care and maintenance as many other mechanical devices do to optimize freezer efficiency, uptime and longevity. Following guidelines and recommended maintenance can ensure proper operation and performance of the ULT freezer.
Unrestricted airflow is critical for proper operation of a ULT.
- Filters and condensers should be regularly inspected for dirt and dust.
- Vacuum the condenser, filters can be vacuumed or washed in mild soap and water then air dried.
Provides energy efficient thermal barrier to keep cold air in and room temperature out. Good gasket seal helps insure product protection from ambient temperature and humidity migration to the interior, reduces frost build up on the inner doors and interior of the unit.
- Seals should be inspected regularly to check for any tears
- Inspect gaskets for deformity and pulling away from frame
- Frost and ice should be removed from around gaskets and between folds of gasket
*Do not use a sharp object that can puncture of dislodge the gasket.
Stay Ahead of Frost and Ice
- Formation of frost is unavoidable
- If not managed the frost will ultimately turn to ice
- Accumulation of ice on gaskets and sealing surfaces can prevent proper sealing
- Buildup of ice on interior walls will insulate the unit, reducing the cooling capacity and increasing recovery times from door openings and product loading
- Regularly wiping off frost from sealing surfaces with a soft cloth, will slow down the formation of ice
- Utilize racks and bins to maximize sample storage, reduce air space and manage sample inventory
- Minimize door opening frequency and time of door opening
- Lack of freezer organization can expose samples to unnecessary temperature fluctuations, excessive stress, limit life of operating components, jeopardizing sample integrity
- Use racks and boxes, bar coded tubes, bar code readers, inventory management systems to locate samples. Quick location of samples, less time freezer doors are open, minimizes frost and stress on ULT
ULT’s should be place on a regular defrost maintenance schedule. We recommend once per year. That can vary depending on daily usage and maintenance practices.
Steps for proper defrost…
- Samples will need to be removed from unit to be defrosted
- Shut down the unit
- Open inner and outer doors, allowing freezer to warm up and ice to melt (24 hours recommended). Place absorbent pads around base of unit to prevent slip hazards
- Unit should be thoroughly dried before placing back into service
- Allow unit to pull down to operating temperature prior to reloading samples (12 hours at setpoint before loading)
Knowledge Center
Top considerations for selecting and maintaining your ULT freezer.
User Manuals
Search for your manual in our Selection Guide for Manuals › |