MicroPlateReadersandWasher
Expand functionality beyond the standard unit
Our reliable and easy to use Thermo Scientific™ Wellwash™ microplate strip washers provide secure washing performance for routine and research ELISA applications. Wellwash washers feature a large color display with graphical user interface, local language versions, and non-pressurized bottles for optimal ease-of-use, convenience, and safety.
Request a quote
Which Wellwash product is best for your needs?
 photo of multidrop combi
Wellwash
photo of multidrop combi nl
Wellwash Versa
Wash head1x8, 1x12two 1x8, two 1x12, 2x8 cell wash head
Wash bottles1 bottle (2 liter)3 bottles (two 4 liter and one 2 liter)
Plate formats9696, 384
Cell washingNoYes
Robot compatibilityNoYes