Create MyQubit .qbt files for the Qubit 3 and 4 Fluorometer
This web tool will help you to create custom MyQubit assay .qbt files that you can upload to your Qubit 3 and 4 Fluorometer. For complete instructions on how to use the MyQubit EZ Guide to generate constants and how to upload .qbt files to your Qubit 3 and 4 Fluorometer, see How to create custom MyQubit assays (Qubit 3.0 and 4).
Please note that this web tool creates .qbt files that can only be used on the Qubit 3 and 4 Fluorometer.
To create .qbt files for the Qubit 2.0 Fluorometer, see Resources for previous Qubit models for detailed instructions.
Calibration
Indicates whether you are creating an assay that requires calibration or not.
"YES" indicates that the assay will require calibration. All assays created using
MyQubit that require calibration will utilize two standards, the first of which is always
a sample blank (analyte concentration of "0") and the second of which is a sample of
a concentration to be determined by the user. "NO" indicates that the assay will not
require calibration. In this case, the output will be a raw fluorescence value or values.
Assay Name
Indicates the name visible on the initial home screen,
i.e., the "family" of assays. For example, "DNA" is
an Assay Name on the factory-configured instrument.
Name Sub-text
Indicates any smaller sub-text under the assay name
(or to the right of the assay name from the graphical
display). For example, "High Sensitivity" is a
Name Sub-text on the factory-configured instrument.
Excitation
BLUE = 470 nm, filter 430–495 nm. RED = 635 nm,
filter 600–645 nm.
Emission
BOTH is only acceptable if Calibration is "NO".
GREEN = 510–580 nm. FAR RED = 665–725 nm.
* Required fields
For Research Use Only. Not for use in diagnostic procedures.