Search
Our TaqMan Assay search tool enables you to explore over 20 million professionally designed ready-to-use assays and arrays, all backed by our TaqMan performance guarantee. New features include access to the latest relevant publications for each assay. Easily navigate the tool to find the best solution for your research. Trusted by researchers, TaqMan Assays have been cited in over 296,000 publications.
Figure 1. A search result like the one shown above will be displayed if the search term applies to multiple assay/array types. The number of relevant assays or arrays will be displayed below the type and will link to the search result set for that type.
5. If desired, refine your search with filters (species, assay attributes, and more) (Figure 2).
Figure 2. Available filters for TaqMan Gene Expression assays.
When searching for TaqMan arrays, you can use any of the following types of keywords:
We offer many tools to help you decide which gene expression assay is right for you:
All TaqMan Assays are designed using our industry-leading bioinformatics design process and meet all of our high-quality-control specifications. For any given target, we compare all assays for that target and select the recommended assay for standard gene expression experiments that detects the maximum number of transcripts for the gene of interest. We also evaluate the assays based on the design criteria below with the recommended assay best meeting the criteria.
For any gene expression assay, details about the assay design and the detected transcripts are provided. Click on the assay ID to view the complete product information
These details include "Assay Design," which shows the location of primers and probes within the transcript's exon structure and is updated regularly (Figure 2).
Click the Assay ID (Figure 3a) to see "More information" about the assay (Figure 3b), including details to help you choose the best assay for your experiment. This section includes:
Figure 3a. Click the Assay ID to see "More information"
Figure 3b. About the assay, including details to help you choose the best assay for your experiment.
You can view the number of publications that cite the assay and a list of those publications in the assay search results (Figure 4). For each assay ID, you can find the following publication-related information:
Filters are always available at the top of the search page to help you refine your search results. For gene expression results, there are multiple parameters to filter on, including cross reactivity.
The Cross Reactivity filter is used to check assays for cross-species reactivity for use in transgenic animal experiments. When you select a species in the Cross Reactivity filter, the list of results will be updated such that any assays that may cross-react with the species selected will be excluded from the results. Any assays that remain in the list will not exhibit species cross-reactivity and thus are suitable to use in a transgenic study.
To order a TaqMan Assay, two pieces of information are required:
The catalog number for a TaqMan Assay is sequence-agnostic and defines other attributes of the assay, including:
The assay ID is a unique identifier for each assay designed either as part of Thermo Fisher Scientific’s comprehensive collection of predesigned assays or as a custom assay using one of our custom assay design tools.
Use the TaqMan Assay search tool to search for an assay (see "Searching for predesigned TaqMan assays and arrays" above).
In the search result, the assay ID is highlighted in bold at the top of the assay result box. The catalog number is listed on the right-hand side under price and will change if you make changes to the dye or size (Figure 1).
There are multiple ways to reorder an assay:
You can export search results by selecting the assays or arrays to be included using the checkboxes (Figure 1). In the box that appears after selecting one or more assays, you have the option to compare up to three products, add all selected items to the cart, export and download them as a list, or send them via email.
The exported files contain detailed information about the assays you have selected. For example, gene expression assay export files will include the following information for each assay ID selected:
If you are exporting information for TaqMan Array products, you will get an xls file with a tab for each product you selected (identified by catalog number or array ID) and the assay layout information for each array.
The TaqMan Assay search tool includes assays and arrays for the following applications:
For Research Use Only. Not for use in diagnostic procedures.