Plate Sealers
Thermo Scientific plate sealers are designed to work with multiple sealing formats included heat seals, adhesive seals and thermoplastic elastomer (TPE) cap mats. Our range of sealing equipment is designed to meet any lab's needs whether they require the high throughput automation compatibility of the ALPS 3000 or the precise sealing parameters for sealing individual plates of the ALPS 50V.
Choose from a range of plate sealing equipment
Whether you require high throughput or high precision heat sealing, or reliable adhesive seals or cap mat Thermo Scientific has a solution for your application
ALPS 3000 Automated Microplate Heat Sealer |
ALPS 50 V-Manual Heat Sealer |
ALPS 30 Manual Heat Sealer |
SuperSealer | |
---|---|---|---|---|
![]() |
![]() |
![]() |
![]() |
|
Seals Plates | X | X | X | X |
Adhesive Seals | X | |||
Heat Seals | X | X | X | |
Cap Mats | X | |||
Individual Sheets | X | X | X | |
Rolls | X | |||
Touch Screen Control | X | |||
Programmable Temperature | X | X | ||
Programmable Timer | X | |||
Automation Compatible | X |
Virtual demo and installation instructions
ALPS 3000 Automated Microplate Heat Sealer
ALPS 50 V-Manual Heat Sealer
Super Sealer
Plate sealing at your fingertips
Plate sealing is important for researchers who wish to protect plate contents either temporarily or longer term, to avoid contamination and/or evaporation. Manual workflows benefit from adhesive seals that can be applied quickly over the surface of the plate. Other short-term options are cap mats or polystyrene lids to protect the well contents.
Resources
Support
Need technical support? Contact our expert team for technical and application support of Laboratory Products