Tube & Vial Capper and Decappers
Thermo Scientific Capper and Decappers provide manual and automated decapping and capping of Thermo Scientific Matrix and Thermo Scientific Nunc Tubes with screw top closures. Capper and Decapper options for Matrix and Nunc tubes offer optimal torque to prevent sample loss. Reduce the risk of repetitive stress injuries (RSI).
For Open Top tubes, we also offer the manual SepraSeal Cap Removal Tool ›
Discover the level of Capper or Decapper you require
The following options are for Matrix and Nunc tubes.
|
Manual |
Automated |
|
Handheld Capper/Decapper | Decapper 500/Decapper 550 | Capit-All Decapper | |
![]() |
![]() |
![]() |
|
Description | The 8-channel Capper/Decapper processes a full rack of Matrix or Nunc tubes in less than one minute, ergonomically. | Automated Capper/Decapper in ANSI racks with Quick Switch Technology for medium-to-high throughput laboratories. | Capit-All is hands-free and Capit-All IS unit is for integration into automated systems. |
Dimension | 25.1cm x 9.5cm x 5cm (9.87 in. x 3.76 in. x 2 in.) |
600mm x 380mm x 440mm (23.6 in. x 15.0 in. x 17.3 in.) |
32.8cm x 36.0cm x 56.0cm (14.2 in. x 12.9 in. x 22 in.) |
Throughput | Low | Medium to high | High |
Speed | 4 seconds for 8 tubes | Less than 3 seconds | Entire rack in 10 seconds |
Channels | 8 channel | 12 channel | 24, 48 & 96 48 channel |
Interface | Projected capacitive Touchscrren |
Graphical user interface Touchscreen |
Four-button user interface |
Barcode Reader | No | 500 - No 550 - Yes |
Integrated IS system |
Data Resource | No | Data Export | Integrated into automated systems |
Compatibility | Matrix or Nunc specific | Matrix or Nunc 2D barcode storage tube |
Matrix or Nunc 2D barcode storage tube |
Ergonomic | Yes | Yes | Yes |
Additions | Stand | None | IS System |
Shop Models |
Capit-All
Capit-All IS
|
Features of the Capit-All IS Integrated System
Capit-All IS System
The Capit-All IS can be operated as a stand-alone benchtop unit. When research needs to be ramped up, the instrument easily can be integrated with most any robotic system. The Capit-All IS can also process over one million tubes before routine service to maximize uptime.
Select a video to learn more
Resources
Support
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.