Sealing-Tape
Nunc Sealing Tapes minimize evaporation and protect samples from contamination and spilling. Adhesive seals are ideal for all assay applications.
  • Wide range of adhesive seals for every assay
  • Adhesive seals effectively seal all microplate formats
  • Pressure-sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
Thermo Scientific Nunc Sealing Tapes
No records were found matching your criteria
SKU Material / Adhesive Application Color Features Temp. Range
232698 Aluminum / Acrylate
  • PCR & storage in 96-well plates
Silver
  • Non-sterile
  • Pierceable
  • RNase / DNase free
-80° to +150°C
232699 Aluminum / Acrylate
  • PCR & storage in 384-well plates
Silver
  • Non-sterile
  • Pierceable
  • RNase / DNase free
-80° to +150°C
232701 Polyolefin / Acrylate
  • Low autofluorescence
  • DMSO compatible
  • Microscopy
Clear
  • Non-sterile
-70° to +100°C
232702 Polyolefin / Acrylate
  • PCR
  • Fluorescence assays
  • Microscopy
Clear
  • Non-sterile
  • RNase / DNase free
-70° to +100°C
235306 Polyethylene / Silicone
  • Repeated piercing
  • Low autofluorescence
Clear
  • Non-sterile
  • Pierceable
-40° to +90°C
235307 Polyolefin / Silicone
  • Q-PCR
  • Microscopy
  • Lowest fluorescence
Clear
  • Non-sterile
-70° to +100°C
236269* Polyester / Acrylate
  • Stiffer than polyolefin
  • General purpose
Clear
  • Non-sterile
-20° to +100°C
236272 Polyester / Acrylate
  • Opaque
  • Luminescence assay
  • Bottom of plate
White
  • Non-sterile
-40° to +80°C
236366 Polyester / Acrylic
  • Cell culture
  • Sterile applications
Clear
  • Sterile
-70° to +100°C
236370+ Polyester / Silicone
  • General purpose
  • Sample protection
Clear
  • Non-sterile
-70° to +100°C
236703 Vinyl / Acrylate
  • For plate top or bottom
  • Photo-sensitive samples
Black
  • Non-sterile
-40° to +80°C
236707 Polyester / Acrylic
  • Convenient roll dispenser
  • General purpose
Clear
  • Non-sterile
  • RNase / DNase free
-20° to +100°C
241205 Rayon / Acrylate
  • Breathable
  • Cell culture
White
  • Sterile
  • Pierceable
-20° to +80°C
249720 Rayon / Acrylate
  • Breathable
White
  • Non-sterile
  • RNase / DNase free
-20° to +80°C
276014 Aluminum / Silicone
  • General use
  • PCR
  • Photo-sensitive samples
Silver
  • Non-sterile
  • Pierceable
  • RNase / DNase free
-80° to +120°C
+ Only available in the Americas
*Not available in the Americas