Vanquish Core HPLC Systems
Eliminate disruptions to your routine analytical work with the Thermo Scientific Vanquish Core HPLC system. No matter which systems you currently use, switching to Vanquish Core is simple and seamless. A full suite of method transfer tools, including tunable gradient delay volume, makes adoption easy. Like all members of our Vanquish product line, Vanquish Core HPLC systems offer hardware precision, detector sensitivity, and simplicity of operation.
Winning in sustainability
The Vanquish Core HPLC system was awarded Best New Sustainable Lab Product of 2025 as part of the Scientists' Choice Awards® presented by Select Science. Vanquish Core HPLC systems are the first liquid chromatography instruments to achieve Platinum ACT Ecolabel 2.0 certification.
Highlights
Simplified method transfer
The unique, tunable gradient delay volume (GDV) and multiple thermostatting modes help refine separations to match the originating system. Users can tune the idle volume without replumbing or requalifying the system. 
Experiment with adjusting the GDV to simplify your method transfer in our interactive demo.
Flexible separations
Thermo Scientific SmartFlow pumping technology includes automatic compensation for changes in eluent compressibility to ensure flow and gradient precision, independent of eluent composition and backpressure. The pump can operate up to 700 bar and is available in binary, quaternary, and dual-gradient options. 
Intelligent system tools
Two optional components are available to improve productivity, ensure compliance, and keep laboratories running smoothly. The Thermo Scientific Vanquish Solvent Monitor tracks solvent levels to prevent dry solvent channels and waste spills. Meanwhile, the touchscreen Vanquish User Interface provides visual monitoring of system status.
Versatility in analyte detection
A variety of detection options ensure you can collect the most information from your separation. Our portfolio includes diode array detection, near-universal charged aerosol detection, fluorescence detection, mass spectrometric detection and more. 
Easy and reliable tool-free connections
The Vanquish Core HPLC system is fitted with Thermo Scientific Viper fingertight fittings, which support near-zero dead volume connections and enable sharper peaks and tool-free setup.
CDS software
The Vanquish Core HPLC system integrates with Thermo Scientific Chromeleon Chromatography Data System (CDS) with compliance tools, networking capabilities, automation, and data processing. We offer standard instrument integration (SII) packages to connect Thermo Scientific HPLC systems to other CDS and MS software programs.
Build your ideal HPLC or UHPLC with our step-by-step configurator
Featuring the ACT Ecolabel
Thermo Scientific Vanquish Core HPLC systems are certified with the prestigious My Green Lab® ACT® Ecolabel awarded by My Green Lab, Inc. The My Green Lab ACT Ecolabel serves as the premier ecolabel for lab products, encompassing consumables, chemicals, and equipment. Standing for Accountability, Consistency, and Transparency (ACT), this certification offers clear, standardized information about a product’s environmental impact across its life cycle covering manufacturing, energy and water use, packaging, and end-of-life disposal. Leverage the My Green Lab ACT Ecolabel to make informed purchasing choices based on transparent environmental data so you can advance sustainability without sacrificing performance.
Videos
System specifications
The table below provides an overview of the Vanquish Core HPLC system specifications for a standard setup. Not all specifications are listed. Your specifications may differ based on the components of your system. Find the specification sheets for the system components in the Component specifications, below.
Category Vanquish Core HPLC Systems
System pressure, maximum 700 bar
Flow range 0–10 mL/min
Pump options (default gradient delay volume)
Binary (400 µL) 
Quaternary (679 µL) 
Isocratic 
Delay volume dependent on pump(s) selected
Biocompatible No
pH range 1–13, chloride concentration up to 0.1 mol/L
System design Modular
Touchscreen user interface display compatible (optional) Yes
Solvent monitoring system compatible (optional) Yes
Customizable gradient delay volume to simplify method transfer  Yes, with method transfer kit
SmartInject technology for maximum injection precision and improved column lifetime Yes
Injection volume range
Standard: 0.01-100 µL
Optional: 0.01-1000 µL
Sample cooling
Yes, 4-40°C
Dependent on sampler selected
Sample capacity (standard)
Based on standard 12 mm OD vials (≤ 1.5 mL)
4 racks of 54 vials (≤ 1.5 mL) each, other rack formats available
+ capacity of 12 × 22.5 mm OD vials (≤ 10 mL) in the carousel
Sampler extension available (optional)
Yes, with Vanquish Charger
Column compartment thermostatting Yes, 5-85°C 
Still air and forced air
Column compartment pre-heater
Active (optional) 
Passive 
Column compartment post-column cooler No
Detector choices
Vanquish Charged Aerosol Detector
Vanquish Diode Array Detector
Vanquish Fluorescence Detector
Vanquish Multiple Wavelength Detector
Vanquish Variable Wavelength Detector
Refractive Index Detector
Mass Spectrometer
Component specifications
Standard Vanquish Core HPLC systems are made up of the following components. Download the associated specifications sheet for more detail on each component.
  Vanquish Core Binary HPLC Vanquish Core Quaternary HPLC Vanquish Core Isocratic HPLC
Pump
Download Specification Sheet
Vanquish Binary Pump C Vanquish Quaternary Pump C/CN Vanquish Isocratic Pump C
Autosampler
Download Specification Sheet
Vanquish Split Sampler C/CT
Extended sample capacity (optional)
Download Specification Sheet
Vanquish Charger (optional)
Column compartment
Download Specification Sheet
Vanquish Column Compartment C
Virtually zero dead volume system connections
Download Specification Sheet
Viper UHPLC Fittings Kits
Detectors
HPLC and UHPLC Detectors page
Optional components: Fraction Collector Visit our Vanquish Fraction Collector FT page
Contact us to get a quote or discuss an HPLC/UHPLC solution
Our general purpose product lines, including LC-MS products, are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks. They are designated for Research Use Only Applications.
For Research Use Only - Not For Use In Diagnostic Procedures.