    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SSEA3 Antibodies
          Invitrogen
          SSEA3 Monoclonal Antibody (MC-631)
          3 Published Figures
          7 References
          View all (10) SSEA3 antibodies
          Questions & Answers
          Cite SSEA3 Monoclonal Antibody (MC-631)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          • Published Figures (3)
          SSEA3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SSEA3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 7
          SSEA3 Antibody (MA1-020) in ICC/IF
          Immunofluorescent analysis of SSEA-3 using anti-SSEA-3 monoclonal antibody (Product # MA1-020) shows expression in human teratocarcinoma NCCIT cells (shown in green) but not in negative control HeLa cells. Formalin fixed cells were permeabilized with 0.1% Triton X-100 in TBS for 10 minutes at room temperature. Cells were blocked with 1% Blocker BSA (Product # 37525) for 15 minutes at room temperature. Cells were probed with a rat monoclonal antibody recognizing SSEA-3 (Product # MA1-020), at a dilution of 1:50 for at least 1 hour at room t... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SSEA3 Antibody in Immunocytochemistry (ICC/IF)
          SSEA3 Antibody in Immunocytochemistry (ICC/IF)
          SSEA3 Antibody in Flow Cytometry (Flow)
          SSEA3 Antibody in Flow Cytometry (Flow)
          SSEA3 Antibody in Immunocytochemistry (ICC/IF)
          SSEA3 Antibody in Immunocytochemistry (ICC/IF)
          SSEA3 Antibody in Flow Cytometry (Flow)
          Product Details
          MA1-020
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:100-1:500
          View 3 publications 3 publications
          Flow Cytometry (Flow)
          1:100
          View 3 publications 3 publications
          Miscellaneous PubMed (Misc)
          -
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Human
          Published species
          Deer, Human, Mammal
          Host/Isotype
          Rat / IgM
          Class
          Monoclonal
          Type
          Antibody
          Clone
          MC-631
          Immunogen
          Fischer rat immunized with 4-8 cell stage mouse embryos
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • DyLight 488
          • DyLight 650
          • PE 
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography - MBP
          Storage buffer
          potassium phosphate with 0.15M NaCl, 1mg/mL BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2536682
          Target Information
          SSEA-3 (stage-specific embryonic antigen 3) is a cell surface antigen, expressed along with SSEA-4, TRA-1-60 and TRA-1-81 in embryonic stem cells, embryonal carcinoma cells, and induced pluripotent stem cells (iPS). In human cells, these surface markers are down-regulated during the differentiation process, while SSEA1 is absent in undifferentiated human stem cells and upregulated on the cell surface after retinoic acid mediated differentiation. Conversely, in mouse stem cells, SSEA-1 expression decreases with differentiation while SSEA-3 and SSEA-4 expression increase.
          Bioinformatics
          Protein Aliases: SSEA-3; stage-specific embryonic antigen 3
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline