    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • P-Glycoprotein (ABCB1) Antibodies
          Invitrogen
          ABCB1 Monoclonal Antibody (UIC2), APC
          Questions & Answers
          Cite ABCB1 Monoclonal Antibody (UIC2), APC
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          ABCB1 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          ABCB1 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          ABCB1 Antibody (MA574202) in Flow
          Flow cytometry analysis of ABCB1 in HEP-G2 cells (red-filled) from human peripheral whole blood cells (black-dashed). Samples were incubated with monoclonal ABCB1 (Product # MA574202) using a dilution of 10 µL reagent per million cells in 100 µL of cell suspension, followed by APC. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ABCB1 Antibody in Flow Cytometry (Flow)
          ABCB1 Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          ABCB1 Monoclonal Antibody (UIC2), APC
          Product Details
          MA574202
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          10 µL/10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          UIC2
          Immunogen
          NIH 3T3 cells transfected with human CD243 (MDR-1) cDNA.
          3D Epitope / Immunogen
          Conjugate
          APC APC APC
          • Unconjugated
          • Alexa Fluor 647 
          • PE 
          • Request custom conjugation
          Excitation/Emission Max
          651/660 nm View spectra spectra
          Form
          Liquid
          Purification
          Size-exclusion chromatography
          Storage buffer
          PBS, pH 7.4, with 0.2% BSA
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is an ATP-dependent drug efflux pump for xenobiotic compounds with broad substrate specificity. It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anticancer drugs. This protein also functions as a transporter in the blood-brain barrier.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: ATP-binding cassette, sub-family B (MDR/TAP), member 1; colchicin sensitivity; doxorubicin resistance; MDR1; P-glycoprotein; multidrug resistance protein 1; multidrug-resistance; P-glycoprotein; P-glycoprotein 1; phospholipid transporter ABCB1; unnamed protein product; uORF
          View more View less
          Gene Aliases: ABC20; CD243; CLCS; ENPAT; GP170; MDR1; p-170; P-GP; PGY1
          View more View less
          UniProt ID: (Human) P08183
          View more View less
          Entrez Gene ID: (Human) 5243
          View more View less
          Function(s)
          protein binding ATP binding xenobiotic-transporting ATPase activity efflux transmembrane transporter activity ATPase activity transmembrane transporter activity ubiquitin protein ligase binding ATPase activity, coupled to transmembrane movement of substances xenobiotic transporter activity carboxylic acid transmembrane transporter activity phosphatidylcholine-translocating ATPase activity phosphatidylethanolamine flippase activity ceramide floppase activity ATPase-coupled intramembrane lipid transporter activity floppase activity phosphatidylethanolamine floppase activity ABC-type transporter activity
          Process(es)
          G2/M transition of mitotic cell cycle xenobiotic metabolic process response to xenobiotic stimulus phospholipid translocation terpenoid transport transmembrane transport transepithelial transport stem cell proliferation ceramide translocation export across plasma membrane transport across blood-brain barrier carboxylic acid transmembrane transport drug transmembrane export drug transport across blood-brain barrier regulation of chloride transport
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-5799cd5976-qbdqk:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0