Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Mouse: 99% identity (118/119 amino acids identical). Human: 98% identity (116/119 amino acids identical) ~60% identity with Neuroligin-1. ~40% identity with Neuroligin-2.
1 µg/mL of MA5-45697 was sufficient for detection of Neuroligin 3 in 20 µg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 110kDa. Does not cross-react with Neuroligin-1, -2, -4 or -4.
This antibody was formerly sold as clone S110-29.
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. Mutations in this gene may be associated with autism and Asperger syndrome. Multiple transcript variants encoding distinct isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ASPGX1; AUTSX1; Gliotactin homolog; neuroligin 3 protein; Neuroligin-3
Gene Aliases: A230085M13Rik; HNL3; KIAA1480; NL3; NLGN3
UniProt ID: (Human) D3DVV0, (Mouse) Q8BXR4, (Rat) Q62889
Entrez Gene ID: (Human) 54413, (Mouse) 245537, (Rat) 171297
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support