Dedicated cryo-FIB microscope to prepare cryo-lamellas for cryo-electron tomography
The Thermo Scientific Aquilos 2 Cryo-FIB is dedicated to the preparation of thin, electron-transparent lamellas for high-resolution cellular cryo-electron tomography or MicroED of micro-crystals.  With the Aquilos 2 Cryo-FIB, you can automate steps in the workflow to produce multiple lamellas from specific programmed locations. The improved dedicated cryo-hardware provides longer run times at ultra-low contamination rates. 
Instrument features
Automated batch cryo-lamella milling
Explore state-of-the-art automation for cryo-lamella production and work with challenging bulk-frozen samples that require cryo-lift-out sample preparation.
Correlative fluorescence and electron imaging in one system
The Thermo Scientific iFLM Correlative System (Integrative Fluorescence Light Microscope) allows you to combine fluorescence imaging and ion milling within a single cryo-FIB SEM microscope.
Aquilos Cryo-FIB resources
Learn more from scientists who are using the Aquilos Cryo-FIB and take a closer look with on-demand webinars and videos.
Cryo-FIB for your lab
Choose the best cryo-focused-ion-beam (cryo-FIB) for your laboratory.  Learn more about other microscope models.
Cryo-ET service and support
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and workflow validation. 
Explore cryo-electron tomography
The Thermo Scientific cryo-electron tomography (cryo-ET) workflow is a complete end-to-end workflow covering cell culture to flash freezing cells to final 3D visualization and analysis.
For Research Use Only. Not for use in diagnostic procedures.