On this page:
A single platform for every phase of your study
Using microarray analysis across agrigenomics segments helps garner stronger insights. This means researchers and scientists can be informed and can confidently make decisions that support plant and animal efforts.
Agrigenomics genotyping solutions from Thermo Fisher Scientific provide breeders and researchers with a powerful and flexible range of genotyping tools to help cost-effectively identify and verify complex genetic traits in plants and animals.
The GeneTitan MC Fast Scan Instrument offers higher throughput and laboratory productivity per technician by helping minimize user intervention and allowing unattended, overnight processing of large numbers of samples in parallel. This unique degree of hands-off automation means your data can be consistently reproducible, your lab can be more productive, and you can spend less time acquiring and managing data and have more time for science.
New SwiftarrayStudio Microarray Analyzer
Coming soon for agrigenomics
GeneTitan MC Fast Scan Instrument
The GeneTitan MC Fast Scan Instrument and proven Axiom array plates in 96- and 384-array layouts are the first to provide a hands-free, automated solution for genome-wide SNP genotyping.
The instrument automates array processing from target hybridization to data generation by combining a hybridization oven, fluidics processing, and an advanced imaging device into a single bench-top instrument.
The imaging device in the GeneTitan MC Fast Scan Instrument uses an external, high-intensity xenon lamp and dual excitation and emission filters to capture images from array plates for use in genotyping and microbiome studies.
Key benefits include:
  • Fast—condenses hands-on processing time to as little as 30 minutes, images an array in less than five minutes, and operates unattended overnight
  • Scalable—meets both medium- and high-throughput needs, enables fast time to data, and requires minimal manual intervention
  • Flexible—supports genotyping studies on multiple array plate formats
  • Accurate—enables high-quality, consistent data by processing multiple samples under identical conditions
  • Adaptable—creates flexible workflows and sample registration via GeneChip Command Console Software
GeneChip Command Console Software
GeneChip Command Console Software provides customization, automation, and integration flexibility along with full-range support:
  • Provides summary of probe cell intensity data (CEL file generation); enables sample and array registration, data management, instrument control, and automatic and manual image gridding
  • Enables flexibility to customize, automate, and integrate workflows with a variety of laboratory information management systems
  • Supports the full range of assays by enabling easy integration with downstream primary analysis applications such as Axiom Analysis Suite and Transcriptome Analysis Console (TAC) Software
Council on Dairy Cattle Breeding Export Tool
Enabling simple reformatting and upload of Axiom array genotype data to the CDCB database
The Council on Dairy Cattle Breeding (CDCB) conducts genetic evaluation of dairy cattle for economically important traits. The CDCB database provides data for use in research and education, including the development of effective procedures for estimating the genetic merit of dairy animals. The CDCB Export Tool is a companion software application to our Axiom Analysis Suite software that enables the export and direct upload of Axiom array genotype data to the CDCB database.
Product specifications
GeneTitan MC Fast Scan Instrument
Dimensions: 55" W x 33” D x 26" H (139.7 cm W x 83.82 cm D x 66 cm H)
Application: Microarray processing
Weight: 147 kg