alternative to g-banding karyotyping to verify genomic integrity and genomic stability of human embryonic stem cell lines
Verify normal karyotype of your hPSCs
The Applied Biosystems KaryoStat and KaryoStat HD Assays provide an alternative to g-banding karyotyping for pluripotent stem cell lines. The assays offer accurate genotyping (sample ID) plus whole-genome coverage for accurate detection of chromosomal abnormalities with results in 3 to 4 days. The assay includes arrays, reagents, and easy-to-use data analysis software for a cost-effective and streamlined analysis of pluripotent stem cell lines. Now researchers won't have to rely on g-banding karyotype service providers.
Need help?
The KaryoStat Assays offer:
  • Cost-effective and accurate detection of chromosomal abnormalities in pluripotent stem cell lines
  • Karyotyping and genotyping (sample ID) with one assay
  • Simple analysis tool that does not require cytogenetics expertise
  • Results in 3 to 4 days
Instrument requirements
KaryoStat and KaryoStat HD Assays are compatible with the Gene Chip Scanner 3000
KaryoStat and KaryoStat HD Arrays are compatible with GeneChip Scanner 3000.
KaryoStat data analysis
KaryoStat and KaryoStat HD have many advantages over g-banding karyotyping
The KaryoStat and KaryoStat HD Assays include access to free downloadable Chromosomal Analysis Suite (ChAS) Software that allows researchers to confirm absence of chromosomal abnormalities in PSC lines.
Download ChAS 3.2
CellModel Services
CellModel Services
Are you lacking the resources or instrumentation? Let us do it for you. Find out more about the KaryoStat and KaryoStat HD Assay Services. 
Request more information
What is the difference between KaryoStat and KaryoStat HD Assay?
KaryoStat Assayhas a resolution similar to g-banding karyotyping and is recommended for most stem cell research applications. The KaryoStat HD Assay provides higher resolution analysis with greater than 99% sensitivity and can reliably detect 25–50 kb copy number changes across the genome at high specificity with single-nucleotide polymorphism (SNP) allelic corroboration.
  KaryoStat Assay KaryoStat HD Assay
Gains >2 Mb >25–50 kb
Losses >1 Mb >25–50 kb
Absence of heterozygosity >5 Mb >1 Mb
SNP Yes Yes
Coverage Low Highest