Search
The assay design could cause later than expected amplification if the assay being used does not detect all of the splice variants for the gene of interest.
Solution:
| Source of Assays | You need to: |
| TaqMan® Gene Expression Assay | Check the alignment of the assay with target sequence. When you search TaqMan® assay, there is an Alignment Map link under each assay ID when search result is returned. The link leads to an alignment map as shown below:
|
| Custom TaqMan® Gene Expression Assay | Submit a template sequence that is common to all of the alternative splicing variants. |
| Assay designed by yourself | Design the primer and probe on a template sequence that is common for all of the alternative splicing variants. |