verifiler-hero-750x225
A new STR chemistry for paternity and kinship testing
Paternity and kinship testing can present challenges in the complexity of claimed relationships and subsequent statistical analyses.
Thermo Fisher Scientific offers a streamlined workflow solution for paternity and kinship testing that helps maximize a laboratory’s efficiency of sample processing and data management. This solution includes the new Applied Biosystems VeriFiler Express PCR amplification chemistry with high discrimination power and direct amplification capability, and Applied Biosystems Converge data analysis software, an all-in-one, modular, browser-based enterprise platform that integrates DNA data management and analysis into a single software product. Converge software identifies and processes Trio Paternity, Trio Maternity, Duo Fatherless, and Duo Motherless kinship tests by automatically creating cases, adding subjects and profiles, running analyses, and generating reports.
ISO-icon-100x100
New: VeriFiler Express PCR Amplification Kit
Offering superior genotyping results from single-source samples, the VeriFiler Express PCR Amplification Kit uses new 6-dye STR Chemistry for paternity kinship analysis. This solution facilitates high throughput and efficient sample-to-profile processing, typically in less than 2 hours.
verifiler-ex-chart-r2
Benefit
Feature
High discrimination power 25-marker multiplex assay that includes two gender markers as well as the highly discriminatory Penta D and Penta E markers for paternity applications
Maximized throughput
Shorter amplification time of 45 min (typically) in direct amplification mode
Increased flexibility
The new chemistry is tested with commonly used substrates such as treated paper, untreated paper and swabs.
Efficient workflow Data results integrated into the new Converge software to centralize storage for ease of access
Optimized chemistry  The new chemistry is validated with multiple sample collection devices such as treated paper, untreated paper, and swabs. It's also designed to work with the most commonly used substrates.
Substrate performance: results show optimal peak heights and clean baselines
Substrate performance
Need help bringing this product online?
Look no further than our Human Identification Professional Services (HPS) team. Since 2007, the team has successfully completed over 400 validation projects worldwide.
Over 20 technical support specialists, each averaging 8 years of real-world forensic experience, provide customers with in-depth training and support on our instruments, chemistries, and software.
Learn more
Resources
Customer support
If you have additional comments or questions, please contact the customer service team via email.
Technical support
If you are a current customer and need technical support, please contact the global service team via email, or phone us at 1-888-821-4HID (4443). Select option #1 for immediate assistance.