Search
Arcturus DNA extraction and RNA isolation kits provide streamlined methods for obtaining high-quality nucleic acids from very small amounts of tissue or cells, whether you are extracting from formalin-fixed, paraffin-embedded (FFPE) tissues or from very few cells (as few as 10 cells).
The Arcturus PicoPure DNA Extraction Kit offers an easy, streamlined genomic extraction procedure that produces PCR-ready DNA. The process enables reliable and reproducible DNA recovery from as few as 10 cells.
Learn more about PicoPure DNA Extraction Kit
The Arcturus PicoPure RNA Isolation Kit is designed to recover high-quality total RNA from fewer than 10 cells―even from a single cell. This kit is designed to provide consistent, efficient RNA recovery without sacrificing quality. Low-volume elution allows you to maximize RNA recovery from small numbers of cells for gene expression analysis.
Learn more about PicoPure Frozen RNA Isolation Kit
The Arcturus Paradise PLUS FFPE RNA Isolation Kit enables unprecedented gene expression analysis of millions of previously inaccessible samples by unlocking RNA from formalin-fixed, paraffin-embedded (FFPE) tissues. Amplify as little as 5 ng of the isolated, fixed RNA using reagents and methods that have been specially optimized to overcome the challenges of cross-linked FFPE templates.
Learn more about Paradise PLUS FFPE RNA Isolation Kit
Arcturus DNA extraction and RNA isolation kits are for research use only, and are not intended for animal or human therapeutic or diagnostic use.