Simple handling and contamination prevention
Explore the potential of these Thermo Scientific CO2 Incubators – designed to be an economical option with advanced features and outstanding contamination prevention options. Each one is backed by high-quality engineering and offers a reliable CO2 incubation chamber for cell culture excellence.  
Minimize the risk of contamination
Midi Model
The Thermo Scientific Midi model has a 40 L capacity. It is our smallest CO2 incubator – designed specifically for cell culture scientists who require a compact culturing workspace to handle small workloads, perform personal use applications, or simply address limited space environments.
Cytoperm 2
The Cytoperm 2 CO2 Incubator meets stringent requirements essential in drug and vaccine research and development. It offers 180 °C high-temperature sterilization, a unique pyrolytic germ barrier in the humidification system that sterilizes water before it enters the chamber, and a reliable air jacketed heating system. A Tri-Gas model with 6 segmented inner doors is also available.
BB 15
Thermo Scientific BB 15 CO₂ Incubators have a simple and modern design, precise temperature and CO₂ control for optimal growth conditions and a powerful humidification system for short recovery times. Designed with contamination prevention in mind, this model is easy-to-clean and can be purchased with a 100% pure copper interior for permanent contamination control.
BBD6220
The Thermo Scientific BBD6220 CO₂ Incubator is ideal for environments that demand the highest levels of sample integrity including cancer research, tissue engineering and immunology. It offers a large inner chamber at 220 L, an external humidity water supply with a continuous thermal decontamination system and 180 °C high-temperature sterilization for confidence in contamination prevention. 
Compare features of the Thermo Scientific CO₂ Incubators
Model name    
Thermo Scientific Midi 40 CO2 Incubator
Cytoperm 2 Gassed CO2 Incubator
Thermo Scientific BBD 6220 CO2 Incubator
Thermo Scientific BB 15 CO2 Incubator
CO2 control
0-20%
0-20%
0-20%
0-20%
Relative humidity
<90% at 37°C
90%, Controllable
60 to 95%, Controllable
≥ 95%
Temperature range
Ambient +5° to 50°C
Ambient +5° to 50°C
Ambient +5° to 50°C
Ambient +18° to 33°C
Oxygen control
--
Optional 30-90% with Tri-Gas model only
Optional 30-90%
--
Sterilization cycle
--
180 °C high temperature sterilization
180 °C high temperature sterilization
--
Chamber dimensions W x H x D
47 cm x 46.5 cm x 59.7 cm
18.5 in. x 18 in. x 23.5 in.
92 cm x 85.5 cm x 77.5 cm
36.22 in. x 33.66 in. x 30.51 in.
92 cm x 88.5 cm x 77.5 cm
36 in. x 34 in. x 31 in.
63.7 cm x 86.7 cm x 76.6 cm
25.1 in. x 34.1 in. x 30.2 in.
Chamber volume
40 L
220 L
220 L
150 L
Stackable
--
--
Optional
--
Chamber material
Stainless steel
Stainless steel
Stainless steel
Stainless steel or 100% pure copper
Voltage, availability
120 V or 230 V, global
230 V, Europe, Middle East, Africa
120 V, Americas
120 V or 230 V, Europe, Middle East, Africa, China
Additional options
Pyrolytic germ barrier to sterilize external humidity water
RS232 data output
RS232 data output
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.