Search
Having difficulties with your experiment?
We are dedicated to your success. Get back on track. View our expert recommendations for commonly encountered problem scenarios.
View the relevant questions below:
Beginning your experiment?
Visit our
Tips for troubleshooting a failed Countess 3/3FL software update:
If the screen to your Countess 3/3FL Automated Cell Counter does not turn on, there could be 2 reasons for this mode of failure:
1) Power Supply malfunction: If the power supply is malfunctioning then the instrument will not receive power indicated by the blue light not turning on.
Here are some troubleshooting suggestions:
2) Countess hardware issue: Once the power supply being the issue is eliminated then it is likely a Countess hardware failure. In this case, the instrument will have to be sent in for repairs or replaced depending on the service contract. Please contact technical support at Technicalservices@thermofisher.com for further assistance.
To resolve connectivity issues between your Countess 3/3FL Automated Cell Counter and your computer, take the following steps:
Step 1: Network compatibility
Ensure that the Wi-Fi dongle can support 5 GHz network as they will help get a stronger signal. This can be verified by matching the back of the Wi-Fi dongle with the image below: 
Step 2: Strength of the network
Ensure that there are enough bars in the Wi-Fi strength icon. Weaker Wi-Fi signal can cause connectivity issues. 
Check if other instruments with Wi-Fi connectivity are also experiencing similar issues.
Test the Wi-Fi connectivity of the Countess instrument with a mobile hotspot. If mobile hot spot is working, then it would indicate issues with the Wi-Fi signal in the lab.
Step 3: USB port malfunction
The Countess has a front and a rear USB port. Try plugging the USB dongle to the other port and see if that resolves the issue. The USB ports are highlighted as #2 and #7 in the image. 
Step 4: Thermo Fisher Cloud set up
Please ensure that the instrument is linked to a Thermo Fisher Connect account. The files will not save without a Thermo Fisher Cloud destination. Please set up an account and then try again to connect.
Step 5: Instrument country
Please make sure that the instrument is set to the correct country 'USA & Others' to save the files correctly. To check the country, go to Settings > Instrument Settings > Cloud region
Step 6: File format
Our R&D team found that when naming the files, if there is an extra space after the file name, the instrument would not be able to save the file, and an error screen will appear. Please ensure that there are no extra spaces after the file name.
Find additional tips, troubleshooting help, and resources within our Cell Counting Support Center.
If your Countess 3 Automated Cell Counter freezes during operation, try power-cycling the instrument. Remove the power cable or flip the On/Off switch several times. This will discharge the capacitors fully. Allow the instrument to remain powered off for 5 minutes. After 5 minutes, plug the instrument back in and reboot. If the above fails to correct the problem, please contact technical support at Technicalservices@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our Cell Counting Support Center.
Instructions for updating the Countess 3/3FL software are below:
1. Go to Countess Software Downloads page.
2. Download the latest version of the Countess 3/3 FL software to a computer.
3. Copy the latest countess software file to a USB drive.
4. Insert the USB drive into the Countess 3/3FL instrument (Ensure the .exe update is not hidden within a folder on the USB, as this will prevent a successful update).
5. Press the Setting icon in the lower right corner of the home screen. 
6. Select "Software Update" from the instrument Settings menu.
7. Select the update location (USB or Cloud) and click Next.
8. Select the "Update" button. The update process should only take a minute or two.
9. The instrument will automatically restart, and the screen will show when the update is complete.
Find additional tips, troubleshooting help, and resources within our Cell Counting Support Center.
To reset your Countess 3 Automated Cell Counter to factory settings, use the following steps:
Step 1: Open the settings menu 
Step 2: Go to 'Instrument settings' 
Step 3: Press 'Reset instrument' button
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Please consult the following link for basic troubleshooting tips: Cell Counting Information for more information. If the above fails to correct the problem, please contact technical support at Technicalservices@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our Cell Counting Support Center.
Troubleshooting information for Countess 3 and 3 FL Automated Cell Counters can be found in the troubleshooting guide:
Countess™ 3 and 3FL Automated Cell Counters: Troubleshooting Guide
Find additional tips, troubleshooting help, and resources within our Cell Counting Support Center.
The most recent version of the Countess™ II or Countess™ II FL software is available here. There will be no more software updates to Countess II/IIFL.
The brightness and size sliders establish the thresholds for the imaging algorithm, allowing users to set both the brightness and size ranges for the sample. If the parameters are at their maximum settings, all brightness levels and sizes are counted, so they cannot be further adjusted. To narrow the brightness or size range for the count, you can move the ends of the slider bar or you can hit the icons at the end of the slider bar to adjust them. To demonstrate this while on the ‘Adjust’ screen, repeatedly tap the ‘dim’ icon on the brightness slider or manually move the bottom end of the slider and watch the green brightness bar decrease in height. The resulting narrowed brightness range can be slid across the brightness scale to include only dim cells, only bright cells, or something in between depending on what is to be included in the count. The same adjustments can be performed for cell size.
Select the “Default” profile to restore the size, brightness, and circularity gates to their maximum.
Uneven illumination on the screen may be fixed by resetting the light cube tray. First, make sure you have the most recent version of the Countess™ II or Countess™ II FL software installed on the instrument (it is available here). If the software is updated, the light cube tray should readjust each time the instrument is power cycled. You can also select the “change light cubes” option under the settings menu, allow the light cubes to reset, and then turn the instrument off and back on. This process may help with uneven screen illumination even if you do not have light cubes present or if you have the Countess™ II instrument (without the fluorescence option).
First, make sure you have the most recent version of the Countess™ II or Countess™ II FL software installed on the instrument (it is available here). You may need to manually adjust the focus and set the nominal focus so that the autofocus will have a set point to focus on the cells. Once cells are stained 1:1 in 0.4% trypan blue and placed in the slide, you may want to let the cells settle for around 30 seconds. Then place the slide in the instrument and after you let the machine autofocus, you may need to refine the brightfield focus with the manual focus. Ideally the live cells should be slightly under focused, where the centers are bright compared to the dead cells that are dark throughout. You may want to zoom in so that you can better see the cells, then manually adjust the focus using the focus slider bar before you hit capture. Once the cells are focused, you can tap the blue “set” box to set the nominal focus; setting the nominal focus will improve autofocus consistency with future slides.
Make sure there are no bubbles or debris visible on the screen that could interfere with the autofocus and make it more difficult to get the sample in the correct focal plane.
You can find some more guidelines for focusing starting on page 20 of the manual.
Decrease the brightfield and fluorescence light intensity before counting the cells.
Once you press “Capture” to count the cells, hit the “Adjust” button and make sure that the size, brightness, and circularity gates are all maximized to include all of the cells in the count. When you go into the “Adjust” screen, you can adjust the size, brightness, and circularity for both live and dead cells, so make sure that you select both the live and dead buttons in the adjust screen so that you are completely maximizing the size and brightness gates for both the live and dead cell population. It’s best to expand the gates first, make sure all of the cells are counted, and then narrow them if you want to exclude cells of a certain size or certain brightness.
You can also select the “Default” user profile to restore the size, brightness, and circularity gates to their maximum.
If the gates are fully maximized, the CSV should indicate 0–60 for cell size min and max and 0–255 for brightness min and max for both live and dead cells.
When you use the test beads, we recommend vortexing the bead stock on high for a full 30 seconds, then pipetting out 10 µL right away to add to 10 µL of trypan blue. The bead and trypan blue mixture should then be pipetted up and down several times to make sure it is well mixed, and the 10 µL sample should be loaded into the slide right away after mixing. The beads can settle quickly, which will affect the concentration. You can also view an image of the beads to ensure that all beads on the Countess™ screen are counted by the counting algorithm.
The light source adjustment controls the LED intensity, camera gain, and exposure time, and it is used for adjusting the image brightness and contrast before you capture the image. Once the cells are inserted, there will be a bar on the right hand side to adjust the light source intensity. Often it helps to turn it down so that the background is slightly darker, which allows better contrast between live and dead cells. Optimal light intensity in brightfield is usually around 10–20 if there are no light cubes in the instrument, and slightly higher around 20–30 if there are light cubes present, but this could vary depending on the sample.
Once the image is captured, you can also adjust the range of cell size and cell brightness that the counting algorithm will count. This doesn’t change the actual brightness of the image or how it looks, but determines which cells will be counted. Once you press “Capture” to count the cells, you can hit the “Adjust” button and make sure that the size, brightness, and circularity gates are all maximized by maximizing the slider bars to include all of the cells in the count. When you go into the “Adjust” screen, you can adjust the size, brightness, and circularity for both live and dead cells, so make sure that you select both the live and dead buttons in the adjust screen so that you are completely maximizing the size and brightness gates for both the live and dead cell population. This is mainly an issue if there are cells that appear in the image, but aren’t being counted. It’s best to expand the gates first, make sure all of the cells are counted, and then narrow them if you want to exclude cells of a certain size or certain brightness.
The brightfield cell concentration assumes that you have diluted the cells 1:1 in trypan blue, so it already takes this dilution into account and multiples the values by 2 so that the cell concentration displayed is the original cell concentration before dilution in trypan blue. The total cell concentration displayed after a fluorescent count does not take any dilution into account, so it is the actual cell concentration in the slide.
Ensure that a FAT-formatted USB drive is inserted in the instrument.
If you are pipetting different samples from the same cell sample, the variability could be due to pipetting or mixing. Use recently calibrated pipettors and make sure that the cell suspension is well mixed by pipetting up and down several times before you add 10 µL to trypan blue. Once the cell sample is mixed with trypan blue, it should also be well mixed by pipetting up and down and loaded into the Countess™ slide right away.
If you are counting replicates of the exact same slide, visually inspect that all cells are counted correctly in the image. Ensure that you do not shake or agitate the slide between counts. There may be a slightly different field of view each time a slide is inserted, so depending on the concentration and uniformity of the cells, there will be some variability when performing replicate counts of the same slide, although it should be less than 10%. When counting fewer cells, a small field of view change for only a small number of cells can have a larger affect, so variability can be reduced by counting cells at a higher concentration.
There are two predominant reasons for this: either the cells in question are truly dead or the focus should be adjusted. In some cases, heavily pigmented cells will also look dead in brightfield; such cells may require fluorescent viability stains.
Ensure that the cells are focused correctly so that live cells have bright centers and dead cells are dark throughout. If the cells are not well focused and look dark on the screen, the Countess™ II or Countess™ II FL instrument will count them as dead cells. If cells are well focused, have bright centers, and are being counted as dead, confirm that they are within the appropriate cell size range and try adjusting the settings. If cells are exposed to trypan blue for a long period of time, viability could be affected so the slide should be prepared and counted fresh each time for best results.
In some cases, heavily pigmented cells will also look dead in brightfield; such cells may require fluorescent viability stains.
To format a USB to FAT32:
Reformatting will result in the loss of all files from the USB drive, so ensure that you have copies of any important files on another drive prior to reformatting. After reformatting, the software should be retransferred onto the USB drive prior to updating the Countess™ II or Countess™ II FL software.
The Countess™ II or Countess™ II FL instrument can also be controlled by a USB mouse, so you can try reinstalling the software by plugging a USB mouse into either USB port on the instrument and use the mouse to perform the software update.
Exposure to light may degrade the dye and these contaminants may promote precipitation. Trypan blue can also form orange/red fibrous aggregates if exposed to refrigeration or freezing temperatures.
Unfortunately, there is no definitive answer to this question. The rate at which the cell-impermeant dye is absorbed depends on the cell type, their state of health, nourishment, engulfment activity, etc.
For Research Use Only. Not for use in diagnostic procedures.