Search
Background fluorescence, sometimes referred to as noise, is the fluorescent signal that you see but you don’t want. Background fluorescence comes from a variety of sources, but it can be separated into two main categories.
Noise from instruments and light sources can be complicated to define and remove, but it tends to remain fairly constant from day to day. Background arising from autofluorescence or from nonspecific staining can be more easily addressed and remedied—at least in theory! Because there can be so many different ways that your experimental design contributes to background fluorescence, you likely have multiple options to reduce it. Some of the sources of background fluorescence in your fluorescent imaging experiment are:
Figure 1. Background fluorescence can arise from many sources in your experiment.
Getting the best data from your fluorescent image requires you to do everything you can to make the difference between your signal (what you want to see fluorescently labeled) and the background (what you don’t want to see fluorescently labeled) as big as possible. That difference is sometimes even expressed as a ratio when you report the quantitative results: ∆F/F, where ∆F is the [signal – background] and F is the background. Of course you want to use a very bright fluorescent dye, but for the best results you also need to have as little background or off-target fluorescence as possible. In general, decreasing background will result in greater image contrast and make it easier to see the desired signal.
Figure 2. Different dye concentrations under identical experimental conditions can produce significantly different amounts of background.
If your sample itself exhibits fluorescence (autofluorescence) that obscures your signal, try labeling with a dye that matches a different filter set (for example, switch from a fluorophore that uses the green filter set to one that uses the red or far-red filter set).
It can be helpful to measure the fluorescence intensity from a well that contains only your cells and the drug or treatment (no fluorescent label) to determine if your treatment is contributing to high background during live-cell imaging. If it is, you may be able to subtract this background value from your results. Alternatively, you may have to choose another treatment or dye that uses a different channel.
The medium that your sample is in can also affect your results. This is true for fixed samples as well as for live-cell imaging. For fixed-cell imaging, most commonly using immunolabeling, you may want to explore mounting media. For live-cell imaging, you will get better signal-to-background if you image in an optically clear buffered saline solution or a medium specially designed to reduce background fluorescence, such as Gibco FluoroBrite DMEM that will also contain the nutrients needed for long-term culture (see live-cell imaging).
The vessel that you image in can also contribute to background fluorescence. For example, plastic-bottom dishes typically used in cell culture can fluoresce very brightly. If you have excess background fluorescence and are imaging in plastic-bottom vessels, try switching to a glass-bottom vessel.
Learn more aboutcell culture vessels designed for imaging
For Research Use Only. Not for use in diagnostic procedures.