Search
The Thermo Scientific ARL X’TRA Companion X-ray Diffractometer is a θ/θ Bragg-Brentano benchtop X-ray diffractometer well suited for Rietveld routine analysis, process control QA/QC, research, and teaching applications.
It is configured with a super solid-state detector to deliver accuracy, precision, safety, and ease of use. As a plus, it comes with a one-click data treatment software to ensure reliable results.
The ARL X’TRA Companion XRD has been developed under the premise that high performance and precision should be associated with ease of use and efficiency, for daily operation with complete peace of mind. Take a look at these benefits.
Equipped with a solid-state pixel detector and advanced photon energy filtering, the system offers fast, high-quality data collection for reliable phase identification, quantification, and crystallinity assessment. The 55 x 55 μm pitch detector helps provide rapid data collection with exceptional energy resolution (<1keV).
We offer diverse sample cups and supports, including zero-background silicon, top-loading, back-loading, and side-loading options. Our specialized supports cater to air-sensitive samples, PMMA, clay, and air filters, helping to ensure optimal performance and accuracy across various research applications.
The ARL X’TRA Companion X-ray Diffractometer offers one-click Rietveld quantification, streamlining data collection to results. An ideal choice for quality assurance, teaching diffraction, or student learning, it helps ensure accurate and reliable results. It supports material science, industrial quality control, and other fields, providing precise results to advance your work.
The ARL X'TRA Companion X-ray Diffractometer supports reflection, transmission, and grazing geometries, making it easy to switch between them. This versatility ensures you have a reliable companion for most of your applications.
The ARL X’TRA Companion X-ray Diffractometer features user-friendly hardware and intuitive software, well suited for teaching powder diffraction. Students quickly become autonomous in their research. The software is compatible with free crystallography databases, offering valuable learning resources.
The ARL X’TRA Companion X-ray Diffractometer helps ensure high accuracy, with measurements validated using NIST standards to within ±0.01° over the full two-theta range.
The ARL X’TRA Companion X-ray Diffractometer offers a resolution of 0.034° two-theta to deliver performance for most of XRD applications. Achieve precise and accurate results with our advanced, compact system.
The ARL X’TRA Companion X-ray Diffractometer's motorized beam knife supports clean data acquisition by reducing sample scattering and automatically optimizing beam alignment, resulting in more accurate and reliable measurements. Start your measurements at 0.3° two-theta and access low-angle data theta with no background interference.
| Goniometer |
|
|---|---|
| Generator |
|
Collimation |
|
| Cooling |
|
| Detector |
|
Sample stage |
|
| Safety |
|
| Software |
|
Achieve precise and efficient XRD analysis effortlessly with the ARL X’TRA Companion X-ray Diffractometer.
From exploration to analysis and processing, either online or in the laboratory, we have everything necessary to meet specifications for quality, environmental responsibility, and productivity. Mining solutions from Thermo Fisher Scientific provide reliable, accurate data and essential information for optimal process control.
The ARL X’TRA Companion X-ray Diffractometer excels in routine cement analysis, offering the flexibility to implement various analytical methods. Its precise phase quantification capabilities ensure minimal errors and consistently reproducible results. The intuitive design enables operators to maintain high standards in quality assurance and control, enhancing overall efficiency in cement production.
The ARL X’TRA Companion X-ray Diffractometer is ideal for teaching and research, featuring open-source software and free databases. It supports various analytical methods, is easy-to-use, robust, and reliable, minimizing the risk of damage even with inexperienced users, and helping to ensure safe operation.
The ARL X’TRA Companion X-ray Diffractometer excels in quality analysis and control, featuring a 6-position sample changer and the flexibility to implement various analytical methods. With Rietveld analysis running seamlessly in the background, it supports precise and consistent results. Its user-friendly design makes it easy for operators to achieve high standards in quality assurance and control.
Explore our comprehensive collection of application notes to see how the ARL X’TRA Companion X-ray Diffractometer can optimize your processes, enhance material characterization, support innovative research, and drive innovation.
For Research Use Only. Not for use in diagnostic procedures.