Search
I am having problems with...
![]() | Your curves are sigmoidal or amplification occurs later than you expected. | |
![]() | You have an amplification product in your no template control (NTC). | |
![]() | You have no detectable PCR product. | |
![]() | If the slope of your standard curve is below -3.6, you have poor PCR efficiency. | |
![]() | Normal Amplification Curve Example Your amplification curve should look something like this |
For Research Use Only. Not for use in diagnostic procedures.