protocol
Quicklinks
Related Product Information
Introduction
The ChargeSwitch® gDNA Purification Kits allow rapid and efficient purification of genomic DNA from small volumes of human blood.  After preparing the lysates, you may purify DNA in less than 15 minutes using the ChargeSwitch® Technology.  Depending on the kit used, samples may be handled individually or in an automated system using a liquid handling robot.
Intended Use for the Kits
The ChargeSwitch® gDNA Blood Kits are designed to allow isolation of genomic DNA from the following amounts of fresh or frozen, human blood treated with the anticoagulant EDTA or citrate.  The purified genomic DNA is suitable for use in downstream applications including PCR, restriction enzyme digestion, and Southern blotting.
  • ChargeSwitch® gDNA 20 µl Blood Kits: Purifies up to 600 ng of genomic DNA from 10-20 µl of human blood.
  • ChargeSwitch® gDNA 100 µl Blood Kit: Purifies up to 3 µg of genomic DNA from 50-100 µl of human blood.
  • ChargeSwitch® gDNA 1 ml Blood Kit: Purifies up to 20 µg of genomic DNA from  1ml of human blood
Important:  Genomic DNA may be isolated from heparin-treated blood; however, the DNA is not suitable for use in downstream applications such as PCR due to the presence of the heparin.
Advantages
Use of the ChargeSwitch® gDNA Blood Kits to isolate genomic DNA provides the following advantages:
Uses a magnetic bead-based technology to isolate genomic DNA without the need for hazardous chemicals, centrifugation, or vacuum manifolds
  • Rapid and efficient purification of genomic DNA from blood samples in less than 15 minutes following sample preparation and lysis
  • Simple lysis with Proteinase K without the need for any mechanical lysis
  • Minimal contamination with RNA
  • The purified genomic DNA demonstrates improved downstream performance in applications including PCR, restriction enzyme digestion, and Southern blotting
  • Includes a kit designed for automated processing of large numbers of 50-100 µl samples in 96-well plates using a liquid handling robot
System Specifications
Starting Material: Varies; optimized for small sample volumes (i.e. 10-20 µl or 50-100 µl or 1 ml)
Elution Volume: 100 µl (20 µl kit), 150 µl (100 µl kit) and 300 µl (1 ml kit)
DNA Yield: Up to 600 ng (20 µl kit), 3 µg (100 µl kit) and 20 µg ( 1 ml kit)
DNA Size: Varies (depends on quality of starting material)
The ChargeSwitch® Technology
The ChargeSwitch® Technology (CST®) is a novel magnetic bead-based technology that provides a switchable surface charge dependent on the pH of the surrounding buffer to facilitate nucleic acid purification.  In low pH conditions, the CST® beads have a positive charge that binds the negatively charged nucleic acid backbone (see figure).  Proteins and other contaminants are not bound and are simply washed away in an aqueous wash buffer.  To elute nucleic acids, the charge on the surface of the bead is neutralized by raising the pH to 8.5 using a low salt elution buffer (see figure below).  Purified DNA elutes instantly into this elution buffer, and is ready for use in downstream applications.  
ChargeSwitch® Magnetic Bead Specifications
Bead Binding Capacity: 5-10 µg genomic DNA per mg
Bead Size: < 1 µm
Bead Concentration: 25 mg/ml
Storage Buffer: 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20
User Supplied Materials
In addition to the reagents supplied with the kit, you need to have the following materials on hand before beginning:
  • A magnetic separation rack suitable for use with 1.5 ml microcentrifuge tubes or 96-well plates
  • Sterile, 1.5 ml microcentrifuge tubes
  • 96 x 2 ml deep well plate (if processing samples in 96-well format; Greiner, Catalog no. 780270, Abgene, Catalog no. AB-0932, or equivalent)
  • 96 x 300 µl U-Bottomed microtiter plate (if processing samples in 96-well format; Greiner, Catalog no. 650201 or equivalent)
  • Vortex mixer
  • 20 µl, 200 µl, and 1 ml sterile, pipette tips
MagnaRack™
The MagnaRack™ (Catalog no. CS15000) is a two-piece magnetic separation rack for use in protocols with magnetic beads, and consists of a magnetic base station and a removable tube rack.  The tube rack can hold up to 24 microcentrifuge tubes.  The tube rack fits onto the magnetic base station in two different positions, associating the row of 12 neodymium magnets with a single row of 12 tubes for simple ‘on the magnet’ and ‘off the magnet’ sample processing.
96-Well Magnetic Separator
The 96-well Magnetic Separator is a magnetic separation rack that can hold up to 96 samples in a deep well plate. The deep well plate fits onto the magnetic base station, associating the array of 24 neodymium magnets with the samples for ‘on the magnet’ and ‘off the magnet’ sample processing (see figures below).
Handling the ChargeSwitch® Magnetic Beads
Follow the guidelines below when handling the ChargeSwitch® magnetic beads.
  • Do not freeze the beads as this irreparably damages them.  Store the beads at room temperature.
  • Always keep the beads in solution.  Do not allow them to dry out as this renders them non-functional.
  • When using the beads, resuspend thoroughly in the storage buffer by vortexing before removal.
  • Discard beads after use.  Do not reuse.
Elution Buffer
ChargeSwitch® Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5) is supplied with the kit for eluting the DNA from the ChargeSwitch® Magnetic Beads.  For best results, use Elution Buffer (E5) to elute the DNA.  Alternatively, TE Buffer, pH 8.5-9.0 is acceptable.  Note that the pH must be between 8.5-9.0 otherwise the DNA will not elute.  Do not use water for elution.
The protocol suggests eluting the genomic DNA in 100 µl (10-20 µl sample) or 150 µl (50-100 µl sample) of ChargeSwitch® Elution Buffer (E5).  You may vary the amount of ChargeSwitch® Elution Buffer (E5) used to obtain genomic DNA in the desired final concentration.  For best results, always use a volume of ChargeSwitch® Elution Buffer (E5) that is equal to or greater than the volume of ChargeSwitch® Magnetic Beads used in the protocol.  If the volume of ChargeSwitch® Elution Buffer (E5) is lower than the volume of beads used, DNA elution is incomplete.  You may need to perform a second elution to recover all DNA.
Automated Liquid Handling
Use of the ChargeSwitch® gDNA 20 µl Blood Kit has been demonstrated on the Tecan Genesis® robotic workstation to purify DNA in a fully automated system from large numbers of 10-20 µl blood samples in a 96-well format.  Other liquid handling robots are suitable provided that each is equipped with a gripper arm, a 96-well magnetic separator, and other additional hardware as described.  This manual provides general guidelines and a protocol that may be used to develop a script for your robot.  Genesis® is a registered trademark of Tecan AG Group.
TOP
Materials
ChargeSwitch® gDNA Blood kit
Product
Number of Purifications
Catalog no.
ChargeSwitch ® gDNA 20 µl Blood Kit
96
960
CS11010
CS11010-10
ChargeSwitch ® gDNA 100 µl Blood Kit
50
CS11000
ChargeSwitch ® gDNA 1 ml Blood Kit
20 CS11001
Shipping and Storage
All components of the ChargeSwitch® gDNA Blood Kits are shipped at room temperature. Upon receipt, store the Proteinase K at 4°C. Store all other components at room temperature.  All components are guaranteed stable for 6 months if stored properly.
Contents
Components
Catalog no.
CS11010
CS11000
CS11010-10
ChargeSwitch ® Lysis Buffer (L12)
100 ml
100 ml
2 x 480 ml
ChargeSwitch ® Magnetic Beads
2 x 1 ml
2 x 1 ml
2 x 10 ml
Proteinase K (20 mg/ml in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl 2, 50% glycerol)
500 µl
500 µl
5 ml
ChargeSwitch ® Purification Buffer (N5)
20 ml
20 ml
100 ml
ChargeSwitch ® Wash Buffer (W12)
50 ml
100 ml
960 ml
ChargeSwitch ® Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)
15 ml
15 ml
100 ml
ChargeSwitch® gDNA 1 ml Blood Kit  (CS11001)
Shipping and Storage
All components of the ChargeSwitch® gDNA 1 ml Blood Kit are shipped at room temperature. Upon receipt, store the Proteinase K at 4°C. Store all other components at room temperature.
All components are guaranteed stable for 6 months if stored properly.
Content
The components supplied in the ChargeSwitch® gDNA 1 ml Blood Kit are listed below. The reagents supplied are sufficient to perform 20 purifications from 1 ml blood samples.
Note:  Some reagents in the kit may be provided in excess of the amount needed.
ChargeSwitch® gDNA Blood kit
Component
Amount
ChargeSwitch ® 10X RBC Lysis Buffer (L8)
25 ml
ChargeSwitch ® WBC Lysis Buffer (L12)
30 ml
ChargeSwitch ® Magnetic Beads
2 x 1 ml
Proteinase K (20 mg/ml in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl 2, 50% glycerol)
500 µl
ChargeSwitch ® Purification Buffer (N5)
4.5 ml
ChargeSwitch ® Wash Buffer (W12)
100 ml
ChargeSwitch ® Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)
10 ml
TOP
Protocol - Purification of Genomic DNA from 10-20 µl Blood Samples
This section provides guidelines and instructions to isolate genomic DNA from 10-20 µl samples of human blood.  Note that the protocol is optimized for efficient purification of DNA from these sample volumes.
Starting Material
Use this procedure to isolate genomic DNA from human blood samples that have been treated as follows:
  • Volume: 10-20 µl of human blood
  • Treatment: EDTA- or citrate-treated
  • Sample state: Fresh or frozen
Before Starting
Perform the following before beginning:
  1. Prepare a Lysis Mix: For each sample, mix 0.5 ml of ChargeSwitch® Lysis Buffer (L12) and 5 µl of Proteinase K to prepare the Lysis Mix.   If you are isolating DNA from multiple samples, you may scale up the volume of reagents used and prepare a master Lysis Mix.
  2. Vortex the tube containing the ChargeSwitch® Magnetic Beads to fully resuspend and evenly distribute the beads in the storage buffer.
  3. Prepare a Purification Mix:  For each sample, mix 20 µl of ChargeSwitch® Magnetic Beads (fully resuspended; see above) and 100 µl of ChargeSwitch® Purification Buffer (N5) to prepare the Purification Mix.  If you are isolating DNA from multiple samples, you may scale up the volume of reagents used and prepare a master Purification Mix.
Preparing the Lysate
Follow the procedure below to prepare a lysate from the 10-20 µl blood sample.
  1. Transfer the 10-20 µl blood sample to a sterile microcentrifuge tube (or a 96 x 2 ml deep well plate).
  2. Add 0.5 ml of Lysis Mix (see above) to the sample and pipet up and down gently 5 times to mix.
    Important:
    Use a 1 ml pipette tip set to 450 µl to mix the sample.  Make sure that the tip is submerged, and pipet up and down gently to avoid forming bubbles.
  3. Incubate the sample at room temperature for 10 minutes or until the sample is clear with no visible lumps.
  4. Proceed to Binding DNA.
Binding DNA
Follow the procedure below to bind the DNA to the ChargeSwitch® Magnetic Beads.
  1. Gently pipet up and down the Purification Mix containing the ChargeSwitch® Magnetic Beads to fully resuspend the beads.
  2. Add 120 µl of ChargeSwitch® Purification Mix to the digested sample (from Step 3, above) and pipet up and down gently 5 times to mix.
    Important: Use a 1 ml pipette tip set to 550 µl to mix the sample. Make sure that the tip is submerged, and pipet up and down gently to avoid forming bubbles.
  3. Incubate at room temperature for 1 minute to allow the DNA to bind to the ChargeSwitch® Magnetic Beads.
  4. Place the sample in the MagnaRack™ (or 96-Well Magnetic Separator if using a 96-well deep well plate) for 1 minute or until the beads have formed a tight pellet.
  5. Without removing the sample from the MagnaRack™, carefully remove the supernatant and discard.  Take care not to disturb the pellet of beads by angling the pipette such that the tip is pointed away from the pellet (see figure).  
  6. Remove the sample containing the pelleted magnetic beads from the MagnaRack™.  There should be no supernatant in the tube.
  7. Add 500 µl of ChargeSwitch® Lysis Buffer (L12; without Proteinase K) to the tube and pipet up and down gently 3 times to mix.  Use a 1 ml pipette tip set to 450 µl.
  8. Add 50 µl of ChargeSwitch® Purification Buffer (N5) and pipet up and down gently 3 times to mix.  Use a 1 ml pipette tip set to 500 µl.
  9. Incubate at room temperature for 1 minute.
  10. Place the sample in the MagnaRack™ (or 96-Well Magnetic Separator if appropriate) for 1 minute or until the beads have formed a tight pellet.
  11. Without removing the sample from the MagnaRack™, carefully remove the supernatant and discard.  Take care not to disturb the pellet of beads by angling the pipette such that the tip is pointed away from the pellet.
  12. Proceed immediately to Washing DNA, below.
Washing DNA
  1. Remove the sample containing the pelleted magnetic beads from the MagnaRack™ (Step 11, above).  There should be no supernatant in the tube.
  2. Add 500 µl of ChargeSwitch® Wash Buffer (W12) to the sample and pipet up and down gently twice to resuspend the magnetic beads.
    Important: Use a 1 ml pipette tip set to 900 µl to mix the sample. Make sure that the tip is submerged, and pipet up and down gently to avoid forming bubbles.
  3. Place the sample in the MagnaRack™ for 1 minute or until the beads have formed a tight pellet.
  4. Without removing the sample from the MagnaRack™, carefully remove the supernatant and discard.  Take care not to disturb the pellet of beads by angling the pipette such that the tip is pointed away from the pellet.
  5. Proceed to Eluting DNA.
Eluting DNA
  1. Remove the sample containing the pelleted magnetic beads from the MagnaRack™ (Step 4, above).  There should be no supernatant in the tube.
  2. Add 100 µl of ChargeSwitch® Elution Buffer (E5) (or TE Buffer, pH 8.5) to the sample and pipet up and down gently 10 times to resuspend the magnetic beads.
    Important: Do not use water for elution. The DNA will not elute due to the poor buffering capacity of water.
  3. Incubate at room temperature for 1 minute.
  4. Place the sample in the MagnaRack™ for 3 minutes or until the beads have formed a tight pellet.
  5. Without removing the tube from the MagnaRack™, carefully remove the supernatant containing the DNA to a sterile microcentrifuge tube (or a 96 x 300 µl U-bottomed microtiter plate).  Take care not to disturb the pellet of beads by angling the pipette such that the tip is pointed away from the pellet.
  6. Discard the used magnetic beads.  Do not reuse the beads.
Storing DNA
Store the purified DNA at -20°C or use immediately for downstream analysis.  Void repeatedly freezing and thawing DNA.
Quantitating DNA Yield
To quantitate the yield of your DNA, we recommend using the Quant-iT™ PicoGreen® dsDNA Quantitation Kit (Catalog no. P7589).  This kit contains the reagents necessary to allow sensitive and accurate fluorescence-based detection of as little as 25 pg/ml of dsDNA using the Quant-iT™ PicoGreen® dsDNA Quantitation Reagent.
TOP
Protocol - Purification of Genomic DNA from 50-100 µl Blood Samples
This section provides guidelines and instructions to isolate genomic DNA from 50-100 µl samples of human blood. Note that the protocol is optimized for efficient purification of DNA from these sample volumes.
Starting Material
Use this procedure to isolate genomic DNA from human blood samples that have been treated as follows:
  • Volume: 50-100 µl of human blood
  • Treatment: EDTA- or citrate-treated
  • Sample state: Fresh or frozen
Before Starting
Perform the following before beginning:
  1. Prepare a Lysis Mix: For each sample, mix 1 ml of ChargeSwitch® Lysis Buffer (L12) and 10 µl of Proteinase K to prepare the Lysis Mix.  If you are isolating DNA from multiple samples, you may scale up the volume of reagents used and prepare a master Lysis Mix.
  2. Vortex the tube containing the ChargeSwitch® Magnetic Beads to fully resuspend and evenly distribute the beads in the storage buffer.
  3. Prepare a Purification Mix:  For each sample, mix 40 µl of ChargeSwitch® Magnetic Beads (fully resuspended; see above) and 200 µl of f you are isolating DNA from multiple samples, you may scale up the volume of reagents used and prepare a master Purification Mix.
Preparing the Lysate
Follow the procedure below to prepare a lysate from the 50-100 µl blood sample.
  1. Transfer the 50-100 µl blood sample to a sterile microcentrifuge tube or a 96 x 2 ml deep well plate.
  2. Add 1 ml of Lysis Mix (see above) to the sample and pipet up and down gently 5 times to mix.
  3. Important: Use a 1 ml pipette tip set to 900 µl to mix the sample. Make sure that the tip is submerged, and pipet up and down gently to avoid forming bubbles.
  4. Incubate the sample at room temperature for 10 minutes or until the sample is clear with no visible lumps.
  5. Proceed to Binding DNA.
Binding DNA
Follow the procedure below to bind the DNA to the ChargeSwitch ® Magnetic Beads.
  1. Gently pipet up and down the Purification Mix containing the ChargeSwitch® Magnetic Beads to fully resuspend the beads.
  2. Add 240 µl of ChargeSwitch® Purification Mix to the digested sample (from Step 3, above) and pipet up and down gently 5 times to mix.
    Important: Use a 1 ml pipette tip set to 900 µl to mix the sample. Make sure that the tip is submerged, and pipet up and down gently to avoid forming bubbles.
  3. Incubate at room temperature for 1 minute to allow the DNA to bind to the ChargeSwitch® Magnetic Beads.
  4. Place the sample in the MagnaRack™ (or 96-Well Magnetic Separator) for 1 minute or until the beads have formed a tight pellet.
  5. Without removing the sample from the MagnaRack™, carefully remove the supernatant and discard.  Take care not to disturb the pellet of beads by angling the pipette such that the tip is pointed away from the pellet (see figure).   
  6. Proceed immediately to Washing DNA.
Washing DNA
  1. Remove the sample containing the pelleted magnetic beads from the MagnaRack™ (Step 5, above).  There should be no supernatant in the tube.
  2. Add 1 ml of ChargeSwitch® Wash Buffer (W12) to the sample and pipet up and down gently twice to resuspend the magnetic beads.  Use a 1 ml pipette tip set to 900 µl to mix.
  3. Place the sample in the MagnaRack™ for 1 minute or until the beads have formed a tight pellet.
  4. Without removing the sample from the MagnaRack™, carefully remove the supernatant and discard.  Take care not to disturb the pellet of beads by angling the pipette such that the tip is pointed away from the pellet.
  5. Remove the sample containing the pelleted magnetic beads from the MagnaRack™.  There should be no supernatant in the tube.
  6. Add 1 ml of ChargeSwitch® Lysis Buffer (L12; without Proteinase K) to the tube and pipet up and down gently 3 times to mix.  Use a 1 ml pipette tip set to 900 µl to mix.
  7. Add 50 µl of ChargeSwitch® Purification Buffer (N5) and pipet up and down gently 3 times to mix.  Use a 1 ml pipette tip set to 900 µl to mix.
  8. Incubate at room temperature for 1 minute.
  9. Place the sample in the MagnaRack™ for 1 minute or until the beads have formed a tight pellet.
  10. Without removing the sample from the MagnaRack™, carefully remove the supernatant and discard.  Take care not to disturb the pellet of beads by angling the pipette such that the tip is pointed away from the pellet.  
  11. Remove the tube containing the pelleted magnetic beads from the MagnaRack™.
  12. Add 1 ml of ChargeSwitch® Wash Buffer (W12) to the tube and pipet up and down gently twice to resuspend the magnetic beads.  Use a 1 ml pipette tip set to 900 µl to mix.
  13. Place the sample in the MagnaRack™ for 1 minute or until the beads have formed a tight pellet.
  14. Without removing the sample from the MagnaRack™, carefully remove the supernatant and discard.  Take care not to disturb the pellet of beads by angling the pipette such that the tip is pointed away from the pellet.
  15. Proceed to Eluting DNA, below.
Eluting DNA
  1. Remove the sample containing the pelleted magnetic beads from the MagnaRack™ (Step 14, above).  There should be no supernatant in the tube.
  2. Add 150 µl of ChargeSwitch® Elution Buffer (E5) (or TE Buffer, pH 8.5) to the tube and pipet up and down gently 10 times to resuspend the magnetic beads.
    Important: Do not use water for elution.  The DNA will not elute due to the poor buffering capacity of water.
  3. Incubate at room temperature for 1 minute.
  4. Place the sample in the MagnaRack™ for 3 minutes or until the beads have formed a tight pellet.
  5. Without removing the tube from the MagnaRack™, carefully remove the supernatant containing the DNA to a sterile microcentrifuge tube (or a 96 x 300 µl U-bottomed microtiter plate).  Take care not to disturb the pellet of beads by angling the pipette such that the tip is pointed away from the pellet.
  6. Discard the used magnetic beads.  Do not reuse the beads.
Storing DNA
Store the purified DNA at -20°C or use immediately for downstream analysis.  Avoid repeatedly freezing and thawing DNA.
Quantitating DNA Yield
To quantitate yield of your DNA, use the Quant-iT™ PicoGreen ® dsDNA Quantitation Kit (Catalog no. P7589).
TOP
Protocol - Purification of Genomic DNA from 1 ml blood
This section provides guidelines and instructions to isolate genomic DNA from 1 ml samples of human blood.
Starting Material
Use this procedure to isolate genomic DNA from:
  • 1 ml of EDTA- or citrate-treated, fresh or frozen, human blood
  • Buffy coats equivalent to 1 ml of white blood cells
Preparing the 1X RBC Lysis Buffer
The first time you use the kit, prepare 1X RBC Lysis Buffer:
  • Mix the contents of the ChargeSwitch® 10X RBC Lysis Buffer (L8; 25 ml) with 225 ml of sterile water to prepare 1X RBC Lysis Buffer (total volume = 250 ml). 
  • Use the 1X RBC Lysis Buffer (see Preparing the Lysate) or store at room temperature.
Preparing the Lysate
Follow the procedure below to prepare a lysate from the 1 ml blood sample.
  1. To a 15 ml centrifuge tube, add the 1 ml blood sample and 10 ml of 1X RBC Lysis Buffer.
  2. Mix by inverting 5 times, then incubate for 5 minutes at room temperature to lyse the red blood cells.
  3. Centrifuge the sample for 5 minutes at 2,000 x g.  Carefully pour away the supernatant, leaving a pellet of white blood cells (visible at the bottom of the tube).
  4. Add 1 ml of ChargeSwitch® Wash Buffer (W12) by dispensing the liquid against the side of the tube.  Take care not to disturb the white blood cell pellet. 
  5. Note: If the pellet is dislodged from the bottom of the tube, centrifuge the sample for 1 minute at 2,000 x g.
  6. Carefully pour away the supernatant containing heme, leaving a pellet of white blood cells.
  7. Shake the bottle of ChargeSwitch® WBC Lysis Buffer (L12) to mix (solution will appear cloudy).  Add 0.5 ml to the sample and mix by vortexing for 10 seconds (recommended) or pipetting up and down 10 times.
  8. Transfer all of the liquid (and any lumps) to a sterile microcentrifuge tube containing 1 ml of ChargeSwitch® WBC Lysis Buffer (L12) and 20 µl of Proteinase K.
  9. Pipet up and down twice to mix.
  10. Incubate the sample for 10-30 minutes at 60°C with occasional mixing (by pipetting up and down, shaking, or vortexing) to lyse the white blood cells.  Do not proceed until the sample is clear with no visible lumps.
  11. Pipet up and down 10 times to thoroughly mix the sample. 
    Note: Use a 1 ml pipette tip and allow as much sample as possible to enter the tip before aspirating the liquid.
  12. Proceed to Binding DNA.
Binding DNA
Follow the procedure below to bind the DNA to the ChargeSwitch® Magnetic Beads.
  1. Vortex the tube containing the ChargeSwitch® Magnetic Beads to fully resuspend and evenly distribute the beads in the storage buffer.  Make sure that all of the solution containing beads is at the bottom of the tube.
  2. Add 100 µl of ChargeSwitch® Magnetic Beads to the digested sample (from Step 10, above), and pipet up and down gently twice to mix.
  3. Add 100 µl of ChargeSwitch® Purification Buffer (N5) to the sample, and pipet up and down gently 5 times to mix.
    Note:
    Adding the ChargeSwitch® Purification Buffer (N5) lowers the pH of the sample, and optimizes the binding conditions.
  4. Incubate at room temperature for 1 minute to allow the DNA to bind to the ChargeSwitch® Magnetic Beads.
  5. Place the sample in the MagnaRack™ for 1 minute or until the beads have formed a tight pellet.  
  6. Without removing the tube from the MagnaRack™, carefully remove the supernatant and discard.
  7. Proceed immediately to Washing DNA.
Washing DNA
  1. Remove the tube containing the pelleted magnetic beads from the MagnaRack™ (Step 6, above).  There should be no supernatant in the tube.
  2. Add 1 ml of ChargeSwitch® Wash Buffer (W12) to the tube and pipet up and down gently 3 times to resuspend the magnetic beads.
    Important: Use a 1 ml pipette tip set to 900 µl to mix the sample. Make sure that the tip is submerged, and pipet up and down gently to avoid forming bubbles.
  3. Place the sample in the MagnaRack™ for 1 minute or until the beads have formed a tight pellet.
  4. Without removing the tube from the MagnaRack™, carefully remove the supernatant and discard.  Take care not to disturb the pellet of beads by angling the pipette such that the tip is pointed away from the pellet.
  5. Repeat Steps 1-4.
  6. Proceed to Eluting DNA.
Eluting DNA
  1. Remove the tube containing the pelleted magnetic beads from the MagnaRack™ (Step 5, above).  There should be no supernatant in the tube.
  2. Add 300 µl of ChargeSwitch® Elution Buffer (E5) (or TE Buffer, pH 8.5) to the tube and pipet up and down gently 10 times to resuspend the magnetic beads.
    Important: Do not use water for elution.  The DNA will not elute due to the poor buffering capacity of water.
  3. Incubate at room temperature for 5 minutes.
    Tip:  For maximum yield, mix the suspension of beads (by pipetting up and down gently) half way through the incubation.  Incubating the sample at 60°C may also improve yield.
  4. Place the sample in the MagnaRack™ for 5 minutes or until the beads have formed a tight pellet.
  5. Without removing the tube from the MagnaRack™, carefully remove the supernatant containing the DNA to a sterile microcentrifuge tube.  Take care not to disturb the pellet of beads by angling the pipette such that the tip is pointed away from the pellet.
    Note: If the eluate containing the DNA is discolored, repeat Steps 4-5.
  6. Discard the used magnetic beads. Do not reuse the beads.
Storing DNA
  • Store the purified DNA at -20°C or use immediately for the desired downstream application.
  • Avoid repeatedly freezing and thawing DNA. Store the purified DNA at 4°C for short-term use or aliquot the DNA and store at -20°C for long-term storage.
Quantitating DNA Yield
You may estimate the yield of purified genomic DNA by checking the UV absorbance at 260 nm or using one of the Quant-iT™ DNA Assay Kits.
UV Absorbance
  1.   Measure the A260 of the solution using a spectrophotometer blanked against 10 mM Tris-HCl, pH 8.5.
  2.   Calculate the amount of DNA using the formula:
DNA (µg) = A260 x 50 µg/(A260 x 1 ml) x dil’n factor x total sample volume (ml)
For DNA, A260 = 1 for a 50 µg/ml solution measured in a cuvette with an optical path length of 1 cm
Quant-iT™ DNA Assay Kits
The Quant-iT™ DNA Assay Kits provide a rapid, sensitive, and accurate method for dsDNA quantitation with minimal interference from RNA, protein, ssDNA (primers), or other common contaminants that affect UV absorbance.  Each kit contains a state-of-the-art quantitation reagent, pre-diluted standards for a standard curve, and a pre-made buffer to allow fluorescence-based DNA quantitation.
TOP
Processing Large Numbers of Samples in 96-Well Format
This section provides general information to use the ChargeSwitch® gDNA 20 µl Blood Kit (Catalog no. CS11010-10) to process large numbers of samples in 96-well format using an automated liquid handling robot.
Hardware Requirements
The ChargeSwitch® chemistry is ideal for purification of DNA using open liquid handling robots, avoiding the need for centrifugation steps or the use of ethanol or chaotropic salts. You will need to have the following hardware to perform automated processing of 10-20 µl blood samples using the ChargeSwitch® gDNA 20 µl Blood Kit:
  • Any liquid handling robotic workstation with a gripper arm
  • Appropriate tips for liquid dispensing and aspiration (see below for factors to consider)
  • 96-Well Magnetic Separator
  • Shaker
  • 96 x 2 ml deep well plate(s) (Greiner, Catalog no. 780270 or Abgene, Catalog no. AB-0932)
  • 96 x 300 µl U-Bottomed microtiter plate (Greiner, Catalog no. 650201)
For an example of how to set up the deck.
Tip Selection
You may use any tips of choice to dispense and aspirate liquid during the purification procedure. Consider the following factors when choosing an appropriate tip to use.
  • Fixed vs. disposable tips
  • Tip size vs. head size
  • Conductive or non-conductive
  • Sterile or non-sterile
  • Filtered or non-filtered
Deck Set Up
Once you have the required hardware, you will need to configure the deck of your liquid handling robot appropriately to process samples. You may use any suitable configuration of your choice. An example is provided below.
Location
Trough Contents
Plate
1
96-well sample tray start position
2
Purification Mix ( i.e. ChargeSwitch ® Purification Buffer (N5) + ChargeSwitch ® Magnetic Beads)
3
Lysis Mix ( i.e. ChargeSwitch ® Lysis Buffer (L12) + Proteinase K)
4
ChargeSwitch ® Purification Buffer (N5)
5
ChargeSwitch ® Lysis Buffer (L12)
6
ChargeSwitch ® Wash Buffer (W12)
7
ChargeSwitch ® Elution Buffer (E5)
8
Waste
9
96-Well Magnetic Separator
10
Shaker
11
96-well Sample Tray
12
ChargeSwitch ® Lysis Buffer (L12)
13
96-well U-bottomed microtiter plate (for final elution)
Primary Liquid Handling Parameters
The table below lists the primary liquid handling parameters required to isolate DNA using the automated protocol. Use the parameters and guidelines provided.
Parameter
Aim
Guidelines
[Magnetic Bead Preparation]
To resuspend beads prior to mixing with solution
  • Only required once
  • Beads stay in suspension for up to 45 minutes
[Mixing #1]
Used to mix beads or bead/DNA pellet with buffer
  • Aspirate/dispense at 400-500 µl
  • Aspirate/dispense position fixed 1-2 mm above the well bottom
  • Use tips/volume setting at 80 µl volume
[Dispense liquid]
Normal liquid parameters for adding a reagent to each well
  • Aspirate/dispense at 300-400 µl. Use multi-dispense if appropriate to save time
[Transfer supernatant to waste]
To remove and discard supernatant
  • Aspirate slowly at 50-100 µl/second
  • Aspirate off the entire liquid volume using liquid detect and tracking or setting fixed height 1 mm above the well bottom
  • Do not disturb pellet
  • Dispense to waste
[Transfer supernatant to another plate]
To transfer supernatant to another plate
  • Aspirate slowly at 50-100 µl/second
  • Aspirate off the entire liquid volume using liquid detect and tracking or setting fixed height 1 mm above the well bottom
  • Do not disturb pellet
  • Dispense slowly at 50-100 µl/second
  • Avoid splashing
[Final DNA Elution]
To dispense the eluate containing DNA
  • Dispense at 10 µl/second
  • Aspirate from position fixed 1 mm above the well bottom
  • Avoid bead carry-over
  • Dispense into new plate at 2 mm above the well bottom
TOP
Automated Genomic DNA Extraction
This section provides a general protocol for automated isolation of genomic DNA from 10-20 µl blood samples in a 96-well format using the ChargeSwitch ®  20µl blood kit (CS11010-10).  Use this general protocol to develop the script for your liquid handling robot.
Materials Needed
  • Liquid handling robot configured to process samples in 96-well plates
  • 10-20 µl blood samples
  • 96 x 2 ml deep well plate(s)
  • 96 x 300 µl U-bottomed microtiter plate
Components Supplied with the Kit
  • ChargeSwitch® Lysis Buffer (L12)
  • Proteinase K
  • ChargeSwitch® Magnetic Beads
  • ChargeSwitch® Purification Buffer (N5)
  • ChargeSwitch® Wash Buffer (W12)
  • ChargeSwitch® Elution Buffer (E5) or TE Buffer (not supplied; 10 mM Tris-HCl, 1 mM EDTA, pH 8.5)
Important Guidelines 
To maximize DNA yield, follow these recommendations when processing your samples:
  • Ensure that the robotic tips enter the wells of the plates without interfering with the pellet of beads.
  • When removing supernatant, leave samples on the 96-Well Magnetic Separator and aspirate slowly to ensure that the pellet of beads is not disturbed. 
  • When resuspending pelleted ChargeSwitch® Magnetic Beads, make sure that all beads are fully resuspended to maximize DNA recovery.
  • To maximize DNA yield, make sure that all Wash Buffer is removed before elution.
  • To maximize DNA yield, make sure that the beads are fully resuspended during the elution step.
Before Starting
Perform the following before beginning:
  • Prepare Lysis Mix: For each sample, mix 0.5 ml of ChargeSwitch® Lysis Buffer (L12) and 5 µl of Proteinase K to prepare the Lysis Mix.  Scale up the volume of reagents used (based on number of samples) to prepare a master mix.
  • Prepare Purification Mix: For each sample, mix 50 µl of ChargeSwitch® Purification Buffer (N5) and 20 µl of ChargeSwitch® Magnetic Beads (make sure that the beads are thoroughly resuspended) to prepare the Purification Mix.  Scale up the volume of reagents used (based on number of samples) to prepare a master mix.
Automated Protocol
Follow the protocol below to isolate genomic DNA from 10-20 µl blood samples. The volumes given are on a per sample basis.
  1. Start with 96 x 10-20 µl blood samples in a 96 x 2 ml deep well plate.
  2. Add 500 µl of Lysis Mix and incubate at room temperature for 10 minutes. Once during the incubation, pipet up and down gently 15 times to mix. Set the pipette tip to 350 µl and avoid forming bubbles.
  3. Add 70 µl of Purification Mix (make sure that the beads are thoroughly resuspended)
  4. Shake at medium fast speed (e.g. pulse, 10 seconds) to evenly distribute the magnetic beads within the solution.
  5. Shake samples rapidly for 20 seconds to mix.
  6. Wait for 30 seconds.
  7. Move samples to the 96-Well Magnetic Separator.
  8. Wait for 90 seconds.
  9. Slowly aspirate all of the supernatant and discard, leaving behind the pellet of beads.
  10. Remove samples from the 96-Well Magnetic Separator.
  11. Add 500 µl of ChargeSwitch® Lysis Buffer (L12; no Proteinase K) and shake samples rapidly for 20 seconds to evenly distribute the magnetic beads within the solution.
  12. Add 50 µl of ChargeSwitch® Purification Buffer (N5) and shake at medium speed for 20 seconds to mix. Samples should appear clear, with no brown flecks.
  13. Wait for 30 seconds.
  14. Move samples to the 96-Well Magnetic Separator.
  15. Wait for 60 seconds.
  16. Slowly aspirate all of the supernatant and discard, leaving behind the pellet of beads.
  17. Remove samples from the 96-Well Magnetic Separator.
  18. Add 500 µl of ChargeSwitch® Wash Buffer (W12).
  19. Shake at medium speed (e.g. pulse, 10 seconds) to evenly distribute the magnetic beads within the solution.
  20. Move samples to the 96-Well Magnetic Separator.
  21. Wait for 60 seconds.
  22. Slowly aspirate all of the supernatant and discard, leaving behind the pellet of beads.
  23. Leave samples on the 96-Well Magnetic Separator for the second wash.
  24. Add 500 µl of ChargeSwitch® Wash Buffer (W12).
  25. Wait for 30-60 seconds.
  26. Slowly aspirate all of the supernatant and discard, leaving behind the pellet of beads.
  27. Move samples to the shaker.
  28. Add 100 µl of Elution Buffer.  Pipet up and down gently 50 times to mix (set the pipette tip to 75 µl).
  29. Shake rapidly for 1-2 minutes to completely disperse the beads within the solution.
  30. Move samples to the 96-Well Magnetic Separator.
  31. Wait for 1 minute.
  32. Slowly aspirate supernatant containing the DNA to a 96 x 300 µl U-bottomed microtiter plate.
Storing DNA
Store the purified DNA at -20°C or use immediately for downstream analysis.  Avoid repeatedly freezing and thawing DNA.
Quantitating DNA Yield
To quantitate yield of your DNA, use the Quant-iT™ PicoGreen ® dsDNA Quantitation Kit (Catalog no. P7589).
TOP
Troubleshooting
Problem
Cause
Solution
Low DNA yield
Incomplete lysis
  • Decrease the amount of starting material used.
  • Be sure to add Proteinase K during lysis.
  • Increase the length of incubation at room temperature.
Insufficient amount of ChargeSwitch® Magnetic Beads added
  • Vortex the tube containing the ChargeSwitch® Magnetic Beads to fully resuspend the beads in solution before preparing the Purification Mix.
  • Before adding Purification Mix to your sample, make sure that the beads are fully resuspended.
Pellet of beads disturbed or lost during binding or washing steps
  • Keep the sample in the MagnaRack or 96-Well Magnetic Separator when removing supernatant during the binding or washing steps.
  • Remove the supernatant without disturbing the pellet of beads by angling the pipette tip away from the pellet.
Bubbles formed during mixing steps
Make sure that the pipette tip is submerged in the solution during mixing.
Incomplete dissociation of DNA from the ChargeSwitch® Magnetic Beads
Perform additional mixing of the suspension of beads (by pipetting up and down).
Low DNA yield, continued
Incorrect elution conditions
  • After adding ChargeSwitch® Elution Buffer (E5) to the sample, pipet up and down to fully resuspend the magnetic beads before incubation.
  • Do not use water to elute DNA. Use ChargeSwitch® Elution Buffer (E5) or TE, pH 8.5.
Lysate mixed too vigorously or small pipette tips used during mixing
  • Use the appropriate pipette tip set to a volume lower than the total volume of solution in the sample.
  • Pipet up and down gently to mix.
No DNA recovered
Water used for elution
Do not use water for elution. The elution buffer must have a pH = 8.5-9.0 or the DNA will remain bound to the ChargeSwitch ® Magnetic Beads. Use Elution Buffer (E5) or TE, pH 8.5.
Added Lysis Buffer containing Proteinase K during second rebinding step
Use ChargeSwitch® Lysis Buffer (L12) without Proteinase K during the second rebinding step.
Didn’t add Purification Buffer during second rebinding step
You must add ChargeSwitch® Purification Buffer (N5) during the second rebinding step to adjust the pH of the solution to allow binding of DNA to the beads.
No DNA recovered, continued
ChargeSwitch® Magnetic Beads stored or handled improperly
  • Store beads at room temperature. Do not freeze the beads as they will become irreparably damaged.
  • Make sure that the beads are in solution at all times and do not become dried. Dried beads are non-functional.
DNA is sheared or degraded
Lysate mixed too vigorously or small pipette tips used during mixing
  • Use a 1 ml pipette tip set to 900 µl to mix the sample.
  • Pipet up and down gently to mix.
Bubbles formed during mixing steps
Make sure that the pipette tip is submerged in the solution during mixing.
DNA repeatedly frozen and thawed
Aliquot DNA and store at 4°C or     -20°C. Avoid repeated freezing and thawing.
DNA contaminated with DNases
Maintain a sterile environment while working ( i.e. wear gloves and use DNase-free reagents).
TOP
LT096