Ion AmpliSeq Exome RDY Kit for Ion GeneStudio S5 Systems
Get rapid whole exome sequencing (WES) results. Go from DNA to variants within 2 days.
Leveraging the ultrahigh-multiplex PCR approach of Ion AmpliSeq technology and the exceptional accuracy of the Ion GeneStudio S5 Systems, the Ion AmpliSeq Exome RDY Kit allows for rapid, accurate sequencing of key exonic regions of the genome, going from DNA to variants in just 2 days.
Test
Discover disease-causing variants in your research with exome sequencing
Exome sequencing is a targeted next-generation sequencing (NGS) approach that is restricted to the protein-coding regions of genomes. The exome is estimated to encompass approximately 1% of the genome, yet contains approximately 85% of disease-causing mutations [1]. For genetic researchers trying to identify the genes implicated in over 6,800 rare diseases [2], exome sequencing enables rapid, cost-effective identification of common single nucleotide variants (SNVs), copy number variations (CNVs), and small insertions or deletions (indels), as well as rare de novo mutations that may explain the heritability of Mendelian and complex disorders [3].
Looking for hybrid capture exome sequencing?
Explore Applied Biosystems hybrid capture solutions for broad, uniform exome coverage using double-stranded DNA probes. Designed for high-throughput sequencing workflows, the DNA Exome solution helps optimize usable sequencing data and support confident variant detection at scale.
Ion AmpliSeq whole exome sequencing NGS workflow
Step 1: Construct library
Ion AmpliSeq Exome RDY Kit
Hands-on time: ~50 minutes
Step 2: Prepare template
Ion Chef System
Hands-on time for Ion Chef System: ~15 minutes
Step 3: Run sequence
Sequencing up to 12 exomes in one day
The Ion GeneStudio Prime System, Ion Chef instruments, and Ion 550 Chip enable sequencing up to 12 exomes per day, 48 per week.*
*Assuming an eight-hour work day and five-day work week. Requires two Ion Chef instruments.
Simple, efficient exome enrichment—see the data
With a workflow requiring less than 50 minutes of hands-on time, the Ion AmpliSeq Exome RDY Kit enables enrichment and library construction in less than 6 hours. The kit requires less hands-on time compared to other exome enrichment technologies (Figure 1), and as little as 50 ng of DNA can be used.
The efficiency of exome enrichment is measured by specificity, defined as the percent of on-target bases, and coverage uniformity, defined as the percent of bases covered at a certain sequencing depth. High uniformity in sequence coverage is important for exome sequencing since this decreases the amount of sequencing needed to achieve a desired coverage threshold (e.g., 20x). Coverage depth is important for accurate variant calling. The Ion AmpliSeq Exome RDY Kit enables high efficiency of enrichment with >90% on-target bases and >90% coverage uniformity (Figure 2).
Figure 1. Comparison of hands-on time and total time for the exome enrichment steps between the Ion AmpliSeq Exome RDY Kit and alternative methods from other vendors.
Figure 2. Comparisons of percentage of bases with 20x coverage and the coverage uniformity between the Ion AmpliSeq Exome RDY Kit and an alternative method from another vendor.
Flexible, on-demand exome sequencing—explore how
Other bench-top sequencing platforms are not as flexible, and running a small number of samples may be costly or researchers may have to wait for large batches of samples to fill each run. With the scalable chip format of the Ion GeneStudio S5 Series, you can cost-efficiently sequence one to four exomes per run without the need to batch samples. Simply select between the Ion 540 and Ion 550 chips (whichever fits your project size). For higher throughput laboratories, sequence up to 48 exomes in a working week* using Ion Chef System, Ion GeneStudio Prime System, and Ion 550 Chip.
Integrated bioinformatics—data analysis in a few clicks
The Ion AmpliSeq exome research solution integrates software and a bioinformatics pipeline to deliver a focused list of annotated exome variants, which helps to reduce the need for expensive bioinformatics resources. 
Torrent Suite Software is preconfigured to support Ion AmpliSeq exome sequencing including optimized variant calling parameters, enabling you to discover relevant variants with confidence. Torrent Suite Software provides a simple run quality report, access to coverage metrics, and full variant calling—all without the need for any command line skills.
Ion Reporter Software enables data to be easily uploaded via a small, efficient file directly from Torrent Suite Software into Ion Reporter Software workflows—allowing researchers access to public annotations of called variants and simple, intuitive filters of only the most relevant variants.
For more complicated experiments, like pairs or trios, more sophisticated Ion Reporter workflows are available to find variants unique to one sample in either a pair or a trio analysis.
Ion AmpliSeq Exome RDY Kit ordering information
Catalog # Name Size Price (USD) Qty
A38262 Each
1,600.00
A38264 Each
1,500.00
For Research Use Only. Not for use in diagnostic procedures.
PMR-003068