Search
Custom and Custom Plus TaqMan™ Copy Number Assays enable the generation of specific, reproducible and easy-to-interpret copy number results. They are suited for validation studies and large sample screens and provide a flexible targeted approach to copy number analysis.
Each custom assay is a mix of forward primer, reverse primer, and FAM™ dye-labeled TaqMan™ MGB™ probe. Designed to run under universal oligo concentration and thermal cycling conditions, these assays are simple to use. Just add TaqMan™ Genotyping Master Mix and your sample to generate sensitive and reproducible data.
There are two custom assay options to meet your research needs (Custom TaqMan® Copy Number Assays and Custom Plus TaqMan® Copy Number Assays). Both allow you to submit a target sequence to our TaqMan® Assay design pipeline, which uses your input sequence to design custom primer and probe sequences. See the key differences between the two options in Table 1:
| Feature | Custom Plus TaqMan® Copy Number Assays | Custom TaqMan® Copy Number Assays | Predesigned TaqMan® Copy Number Assays |
|---|---|---|---|
| Designed with copy number-specific algorithm optimized for performance | ✔ | ✔ | ✔ |
| Available limited to human and mouse assays | ✔ | ✔ | |
| Contains TaqMan® FAM™ dye-labeled MGB™ probes and 2 unlabeled PCR primers | ✔ | ✔ | ✔ |
| Targets undergo SNP and repetitive sequencing mask | ✔ | ✔ | |
| Genome-specific check | ✔ | ✔ | |
| Reference assay compatibility check | ✔ (optional) | ✔ (optional) | ✔ |
| Assay sequences provided | ✔ | ||
| Assay context sequences and genome location provided | ✔ | ✔ |
Browse the GeneAssist™ Copy Number Assay Workflow Builder. Navigate to the human or mouse genomic region of interest to select predesigned assays, or select a targe region for Custom Plus Assay design. Alternatively, submit pre-masked sequences for standard Custom Assay design or submit primer/probe pair sequences for assay synthesis.
| Catalog Number | Product Name | No. of 10 µL reactions (384-well plate) | No. of 20 µL reactions (96-well plate) |
|---|---|---|---|
| 4442487 | Custom Plus TaqMan® Copy Number Assay, SM | 720 | 360 |
| 4442520 | Custom Plus TaqMan® Copy Number Assay, MED | 1,500 | 750 |
| 4442488 | Custom Plus TaqMan® Copy Number Assay, LG | 5,800 | 2,900 |
| 4400294 | Custom TaqMan® Copy Number Assay, SM | 720 | 360 |
| 4400295 | Custom TaqMan® Copy Number Assay, MED | 1,500 | 750 |
| 4400296 | Custom TaqMan® Copy Number Assay, LG | 5,800 | 2,900 |
Approximate delivery times are 4-6 days in the US and 6-10 days in Europe.