Search
Serum-free supplement specifically formulated for growth and expansion of human and rat neural stem cells (NSCs) and progenitor cells (PCs)
Table 1. Components required for NSC culture
| Product | Stock concentration | Final concentration | For 100 mL | For 500 mL |
|---|---|---|---|---|
| StemPro Neural Supplement | 50x | 1x | 2 mL | 10 mL |
| KnockOut DMEM/F-12 | 1x | 1x | 97 mL | 485 mL |
| GlutaMAX-I Supplement | 100x | 1x | 1 mL | 5 mL |
| bFGF | 20 ug/mL | 20 ng/mL | 100 μL | 500 μL |
| EGF | 10 ug/mL | 10 ng/mL | 100 μL | 500 μL |
StemPro Neural Supplement enables superior expansion of GPCs, resulting in a more than 50-fold increase in cell number (Figure 1).
![]() | Figure 1. GPCs were cultured in medium supplemented with StemPro Neural Supplement. Every 7 days, cells were harvested and replated for further proliferation and accumulated cell number was documented. After passage 3, accumulated cell number was more than 50-fold higher than starting cell numbers. |
Compared to GPCs cultured in StemPro Neural Supplement (Figure 2A), GPCs cultured in ScienCell™ OPC medium (a competitor) do not passage further and lose their progenitor state to differentiate into astrocytes (Figure 2B).
Figure 2. GPCs were cultured either in (A) Gibco medium with StemPro Neural Supplement or (B) ScienCell™ OPC medium. During the 3 week culture period, cells cultured in (A) Gibco medium could be passaged 3 times resulting in 50-fold more cells while retaining their phenotype. However, cells cultured in (B) ScienCell™ OPC medium (a competitor) did not increase in number but resulted in a loss of normal phenotype and differentiation to downstream lineages such as astrocytes (large cell body).
Table 2. Components required to culture GPCs
| Product | Stock concentration | Final concentration | For 100 mL | For 500 mL |
|---|---|---|---|---|
| StemPro Neural Supplement | 50x | 1x | 2x | 10 mL |
| KnockOut DMEM/F-12 | 1x | 1x | 97 mL | 485 mL |
| GlutaMAX-I Supplement | 100x | 1x | 1 mL | 5 mL |
| bFGF | 10 μg/mL | 10 ng/mL | 100 μL | 500 μL |
| PDGF-AA | 10 μg/mL | 10 ng/mL | 97 mL | 485 mL |
Human GPCs cultured in StemPro Neural Supplement retain their undifferentiated state as demonstrated by phenotypic marker expression of PDGFRa, Olig2 and N2 markers (Figure 3).
Figure 3. Phenotype marker expression of the human GPCs cultured in medium supplemented with StemPro Neural Supplement. GPCs were labeled with GPC biomarkers such as PDGFRa, Olig2 and NG2. The data reveal the ability of StemPro Neural Supplement to support GPCs to retain their undifferentiated status.
GPCs grown in StemPro Neural Supplement retain their differentiation potential into oligodendrocytes and astrocytes (Figure 4).
Figure 4. Human GPCs were proliferated in the medium supplemented with StemPro Neural Supplement then differentiated further to (A) oligodendrocytes and to (B) astrocytes. Oligodendrocytes were stained with biomarker of GalC (green) and astrocytes were stained with biomarker of GFAP (Red). All nuclei were labeled with DAPI.
1. Is StemPro Neural Supplement serum free?
Yes
2. What are the intended uses of StemPro Neural Supplement?
StemPro Neural Supplement is an optimized serum-free additive for the growth and expansion of neural stem and progenitor cells. For research use only.
3. Do I need to add other supplements?
Mixing of StemPro Neural Supplement with a basal medium such as Knockout DMEM/F-12 or Neurobasal medium and the addition of appropriate quantities of recommended growth factors yields a complete medium.
4. What are the key applications of StemPro Neural Supplement?
StemPro Neural Supplement supports the growth and expansion of neural stem cells and progenitor cells. It also supports the survival of neurons and differentiation of progenitor cells to astrocytes, oligodendrocytes, and Neurons. Besides its application can also be extended to transplantation studies and screening of drug candidates.
5. What species can I use StemPro Neural Supplement to culture?
It can be used for human and rat NSCs, GPCs, and some neuronal progenitors.
Stem Cell Culture Support Center
Relevant technical information, tips and tricks, and answers to everyday problems.