Search
Developing stable cell lines is a vital process in many research applications, including those for studies of long-term genetic regulation, sustained expression in gene therapy, and large-scale protein production in biotechnological and biopharmaceutical settings. Selecting the appropriate antibiotics plays a crucial role in ensuring success, as they offer effective solutions for various research needs; including dual selection and the rapid establishment of stable cell lines. This guide will walk you through the essential steps and considerations for successful stable cell line generation, from the initial selection of antibiotics to the final isolation of antibiotic-resistant clones.
There are many selection antibiotics to choose from for stable cell line development. The antibiotic chosen will depend on which antibiotic resistance gene or selectable marker is used in the transfection experiment.
Thermo Fisher Scientific offers high-quality selection reagents to complement our wide variety of selectable eukaryotic expression vectors. Geneticin (G418 sulfate), zeocin, hygromycin B, puromycin, and blasticidin antibiotics are the most common selection antibiotics used for stable cell transfection. These antibiotics provide unique solutions for your research needs, such as dual selection and rapid, stable cell line establishment.
Selection of stably transfected cells begins with successful transfection of a plasmid containing a selectable marker, such as an antibiotic resistance gene. As a negative control, cells should be transfected using DNA that does not contain the selectable marker.
It is important to note that a kill curve should be established for each cell type and each time a new lot of the selective antibiotic is used. Follow the steps below to generate a kill curve:
Transfect the cells using the desired transfection method. If the selectable marker is on a separate vector, use a 5:1 to 10:1 molar ratio of plasmid containing the gene of interest to plasmid containing the selectable marker.
Note: Perform control transfections with a vector containing the selectable marker but not the gene of interest. If colonies are obtained from cells transfected with the control plasmid but not from cells transfected with plasmid containing the gene of interest, indicating that the gene of interest may be toxic. It is also important to perform replicate transfections in case the transfection fails.
Forty-eight hours after transfection, passage the cells at several different dilutions (e.g., 1:100, 1:500) in medium containing the appropriate selection drug. For effective selection, cells should be sub confluent, because confluent, non-growing cells are resistant to the effects of antibiotics like geneticin. Suspension cells can be selected in soft agar or in 96-well plates for single-cell cloning. For the next two weeks, replace the drug-containing medium every 3 to 4 days (or as needed).
Note: High cell densities in suspension cultures require frequent medium changes that may deplete critical soluble growth factors, thereby reducing cell viability and the efficiency of the system.
During the second week, monitor cells for distinct “islands” of surviving cells. Depending on the cell type, drug-resistant clones will appear in 2–5 weeks. Cell death should occur after 3–9 days in cultures transfected with the negative control plasmid.
Isolate large (500–1,000 cells), healthy colonies using cloning cylinders or sterile toothpicks, and continue to maintain cultures in medium containing the appropriate drug (for the isolation of clones in suspension culture, see Freshney, Culture of Animal Cells: A Manual of Basic Technique and Specialized Applications, Wiley-Blackwell, New York).
Transfer single cells from resistant colonies into the wells of 96-well plates to confirm that they can yield antibiotic-resistant colonies. Ensure that only one cell is present per well after the transfer.
Find products, citations, and protocols optimized for your transfection experiments. Input information on your experiment type, cell line, and payload to unlock solutions.
A stable cell line is a population of cells that have been genetically modified to express a specific gene of interest consistently over many generations. These cells maintain the introduced genetic material without losing it, making them invaluable for long-term studies and protein production.
To generate stable cell lines, transfect cells with a vector carrying the gene of interest and a selectable marker. After transfection, apply selection pressure to eliminate non-transfected cells and isolate colonies that stably express the gene. Expand these colonies to establish a uniform cell line.
Stable cell lines have integrated transfected DNA into the cell's genome, allowing for long term gene expression as the genetic material is passed on during cell division. Transient cell lines only express the transfected genes temporarily and do not integrate them into the cell's genome. In addition, stable cell lines require the use of selection markers and antibiotics to isolate cells with the integrated gene, while transient cell lines do not but work well for short-term studies like functional assays and quick protein expression.
Depending on the specific protocols and the efficiency of each step, generating a stable cell line can take anywhere from a few weeks to several months.
Selection occurs approximately forty-eight hours after transfection, at which point the cells are selected and maintained for the next 2 weeks. Cell death of non-resistant cells should occur after 3–9 days, and based on the cell type, drug-resistant clones can appear in 2–5 weeks. Verifying successful integration and expression can take an additional 1–2 weeks. Always refer to your product inserts for precise information.
For Research Use Only. Not for use in diagnostic procedures.