Search
Quickly and accurately inoculate microtiter plates with enhanced ergonomics using the Thermo Scientific Sensititre 8-Channel Programmable Pipette.
The 8-Channel Programmable Pipette shares common features with all components of the Sensititre AST System: It produces accurate, reliable, consistent results; it fits seamlessly into an integrated AST workflow including manual, semi-, and fully automated options; and customers receive the benefit of premiere Thermo Scientific support and service.
In addition, the Programmable Pipette offers these additional features for optimized inoculation.
Enhanced ergonomics
Keeps your hand in a neutral position, increasing comfort and accuracy. Upright design prevents tips from contamination by touching the bench.
Simple operation
Tip ejection is effortless, and onboard calibration software eliminates the need for any tools. Requires only a 90-minute charge every 600 cycles.
Versatile dispensing options
Delivers 25 to 1,250 µL in 5 µL increment settings, using one to eight tips. Touchpad makes it easy to set up a single delivery or adjust multiple programmable settings. Dispensing speed option allows dosing an entire plate in less than 10 seconds.
Technical characteristics |
Accuracy: ±0.8% at 1250 µL, ±1% at 625 µL, ±3% at 125 µL |
Volume (metric): 25–1250 µL |
|
Increments: 5 µL |
|
Number of channels: 8 |
|
Display: LCD |
|
Physical and electrical characteristics |
Instrument dimensions: 15.24 cm (H) x 20.32 cm (W) x 20.32 cm (D) 6 in (H) x 8 in (W) x 8 in (D) |
Electrical requirements: Rechargeable Lithium-ion Battery, Ovation Power Supply |
|
Includes: Power supply with adapter; shelf hanger |
|
Model: Right-handed |
|
Finger style: Ergonomic |
* Not all products are available for sale in all territories. Please inquire.