Search
The Thermo Scientific Aquilos 2 Cryo-FIB is dedicated to the preparation of thin, electron-transparent lamellas for high-resolution cellular cryo-electron tomography or MicroED of micro-crystals. With the Aquilos 2 Cryo-FIB, you can automate steps in the workflow to produce multiple lamellas from specific programmed locations. The improved dedicated cryo-hardware provides longer run times at ultra-low contamination rates.
For Research Use Only. Not for use in diagnostic procedures.