Search
Modern laboratories need to optimize limited space and maximize analysis time. That means being as efficient as possible with sample handling, sample preparation, and sample transport.
Thermo Scientific SureSTART WebSeal well plates and sealing mats are the perfect solution when there is a need for high throughput and high efficiency in the lab.
Because experiments, analytes, and budgets can vary, we have organized our WebSeal well plates and mats into three performance levels to provide you with the affordability, compatibility, and performance you need.
No HPLC/UHPLC or GC instrument has been left behind. Our well plates are suitable for all vendors' instruments. All WebSeal products are Specification Certified. In addition, well plates are chromatography or LC-MS/GC-MS tested for solvent compatibility and inertness.
Get a sure start with these SureSTART WebSeal products to obtain the performance, reproducibility, and accuracy of results you are looking for. See how our well plate and sealing mat collection delivers a first-class product experience from purchase to final result.
Looking for a vial? Explore our range of SureSTART chromatography autosampler vials and closures.
Select the right SureSTART vial and well plate for your application or instrument type.
| Assured quality and performance | Instrument compatibility | Sample security | Better product experience |
|---|---|---|---|
|
We offer a comprehensive portfolio of WebSeal well plates, sealing mats, kits, and accessories for all of your chromatography needs. Our well plates are designed for instant compatibility with your autosampler and eliminate the hassle of setting up autosampler vials in a vial tray, saving time in your workflow. WebSeal well plates come in a wide variety of well volumes, well shapes, and high-recovery cavity styles so you can choose the plate that meets your analytical requirements. The diagram below illustrates the different well shapes and cavity styles available across the WebSeal well plate product line.
If you are working in a non-regulated laboratory and using water and polar solvents, our Performance Level 1 plastic, non-coated, non-sterile, chromatography tested well plates are the perfect solution for you.
| Level 1: 384-well plates for everyday applications | ||
|---|---|---|
![]() | 384-Well Microtiter Plates, Square V-Bottom | 384-Well Mid-well Plates, Square V-Bottom |
| Compatible sealing mats | ||
![]() | 384-Well Plate Sealing Mat, Square, Level 1 | 384-Well Plate Sealing Mat, Square, Level 1 |
Select Performance Level 2 plastic, non-coated, non-sterile, GC-MS and LC-MS-tested well plates when working in a regulated laboratory that uses water and polar solvents.
| Level 2: 384-well plates for high-throughput applications | ||
|---|---|---|
![]() | 384-Well Microtiter Plates, Square U-Bottom | 384-Well Mid-well Plates, Square V-Bottom |
| Compatible sealing mats | ||
![]() | 384-Well Plate Sealing Mat, Square, Level 2 | 384-Well Plate Sealing Mat, Square, Level 2 |
If you need to meet laboratory regulations and use all solvent types including non-polar solvents for your chromatography applications, our unique Performance Level 3 WebSeal Plate+ glass-coated plastic plates are ideal for your workflows. We also offer convenient, pre-assembled well plate kits along with accessories and replacement parts that provide the efficiency you need with the high-quality materials you expect from Level 3 products.
| Level 3: Plate+ 384-well plates for high performance applications | ||
|---|---|---|
![]() | Plate+ 384-Well Microtiter Plates, Square-Rounded V-Bottom | Plate+ 384-Well Mid-well Plates, Square-Rounded V-Bottom |
| Compatible sealing mats | ||
![]() | 384-Well Plate Sealing Mat, Square, Level 1 384-Well Plate Sealing Mat, Square, Level 2 | 384-Well Plate Sealing Mat, Square, Level 1 384-Well Plate Sealing Mat, Square, Level 2 |
| Level 3: well plate kits for high performance applications | |
|---|---|
![]() | 96-Well Plate Kits with Sealing Mat and Inserts |
![]() | 96-Well Plate Kits with Inserts and Snap Caps/Plugs |
| Level 3: well plate kit accessories and replacement parts | ||||
|---|---|---|---|---|
| Well Plate Vials and Inserts for 96-Well Plates | Caps for 96-Well Plate Vials and Inserts | 96-Well Silicone/PTFE Mats | 96-Well Plates for Well Plate Kits | Micro-Tube-Rack-System, 96 Positions |
With the SureSTART WebSeal product line, you gain all the benefits of a single well plate and sealing mat portfolio that is tailored for chromatography and divided into three distinct performance levels to meet all of your scientific and budget needs. Selecting the perfect WebSeal plate and mat for your chromatography requirements has never been easier with the following selector tools and resources below.
* Required field