Sensititre SWIN Epidemiology Module
Comprehensive AST reporting to track resistance and support stewardship
Comprehensive AST reporting to track resistance and support stewardship
Track and monitor regional antibiotic resistance trends and compile antibiograms with the comprehensive reporting options in the SWIN Software Epidemiology Module. This module makes it easy to generate complete, real-time reports and bar graphs to support your antimicrobial stewardship program and surveillance of antimicrobial resistance patterns.
Features
The SWIN Epidemiology Module allows users to access data quickly via detailed reporting capabilities and multiple search options. In just minutes, laboratories can create comprehensive reports of geographically distinct resistance patterns and track changes over time as part of their antimicrobial susceptibility testing and stewardship programs.
Create and generate comprehensive reports
Without manual data calculations or expensive interface customization.
Track susceptibility trends
Accurately and easily.
Customized report options
Percent Susceptible
(Routine Antibiogram/JCAHO requirement)
Cumulative MIC
Workload Report
Occurrence Report
Interpretation Report
The SWIN Epidemiology Module
Provides comprehensive reporting of AST results to help detect, monitor, and investigate local antibiotic resistance patterns.
Ordering information
* Not all products are available for sale in all territories. Please inquire.