Search
Select the level of automation your ion chromatography (IC) application requires from a range of Thermo Scientific Dionex IC autosamplers, which include high-precision low-cost, metal-free, and high-sample-volume autosamplers and manual injectors.
Browse our complete portfolio below, or let one of our experts help you tailor a selection specific to your application and workflow needs.
Dionex AS-AP Autosampler | Dionex AS-DV Autosampler | Dionex AS-HV Autosampler | |
|---|---|---|---|
![]() | ![]() | ![]() | |
| Request quote | Request quote | Request quote | |
| Features | High precision, reliability, ruggedness, and ease of use; automates many bench-top tasks; can be used with entire ICS product family | A low-cost, metal-free, rugged, automated sample loading solution with random access and sample preparation capabilities | High sample volumes up to 250 mL; AutoPrep option for trace analysis; compact footprint |
| Applications | Simultaneous and sequential injection, concentration, matrix elimination, fraction collection and sample conductivity/pH measurement | Simultaneous injection, concentration, matrix elimination | Simultaneous injection, concentration, matrix elimination |
| Injection | Type: Electrically actuated Rheodyne valves | Volume: 0.1–5.0mL in 0.1mL increments | Type: Full loop/concentrator |
| Dispense Speed | -- | 0.1–5.0mL/min in 0.1mL/min increments | -- |
| Syringe Size | 100, 250, 1000, 5000μL | -- | -- |
| Vial Size | 10 mL polystyrene vials 1.5 mL glass vials 0.3 and 1.5 mL polypropylene vials 0.10, 0.45, 0.5, and 2.0 mL well plates | -- | 10, 15, 26, 40, 50, 60mL vials |
| Sample Capacity | 10mL vials—81 samples 0.3 or 1.5mL vials—120 samples 0.45, 0.5, or 2mL (three 96 well plates) 1152 samples (three 384 well plates) | 50 PolyVials 0.5mL, 5.0mL, or mix of the two | 63 × 50mL: 72 × 26mL; 120 × 15mL, 180 × 10mL, and 63 × 40/60mL ASE vials |
Get reliable performance and productivity with quality parts and accessories designed for your IC autosampler.