Search
Address your challenging charge variant separations by using the Thermo Scientific ProPac Elite WCX 5µmLC columns. This weak cation exchange (WCX) column builds on popular ProPac WCX-10 columns by reducing the particle size of the nonporous resin from 10μm to 5μm, thus providing even faster and higher efficiency and resolution separations of proteins and glycoproteins.
ProPac Elite columns work well with pH gradients. It’s a simpler method for mAb analysis.
You need to monitor the charge variant profiles of mAbs closely. Learn how the newest addition to our comprehensive portfolio of biopharma columns help you take control by delivering high resolution data and consistent column to column performance.
It’s easy to produce linear pH gradients for fast and robust ion exchange separations of your therapeutic protein. Learn how patented pH gradient buffer kits combined with IEX chromatographic columns enable you to...
This brochure celebrates our rich history in Bio LC columns, highlights key product features and provides example applications demonstrating exceptional column performance.
Need more information about the ProPac Elite WCX columns and how it enhances your charge variant separations?