Search
Gas cluster ion sources permit XPS depth profiling of soft materials without perturbing the chemistry of the remaining material, enabling you to measure and analyze more materials that were previously difficult to characterize with a standard ion beam.
Learn from our XPS experts:
From investigating self-assembled monolayers to bio-sensors and implants, surface analysis reveals critical information on both the chemistry and conformity of materials that have been designed to mimic biological systems.
Discover how X-ray photoelectron spectroscopy can be used to investigate the surfaces of a variety of samples in the growing field of biomaterials.
This webinar may interest:
Our webinar presentation shows how a multi-technique approach using both Raman spectroscopy and XPS addresses the challenges posed at all steps in the creation of new graphene devices – from guiding the initial graphene synthesis, transfer to the desired substrate, and understanding chemical modification and analysis of the finished device.
A vibrational spectroscopy, the very high sensitivity of Raman makes it an ideal probe for a number of important properties specific to graphene and nanotube samples, such as layer thickness, sample integrity, and tube diameter.
X-ray photoelectron spectroscopy (XPS) is ideally suited to the determination of the surface chemistry and the way in which that chemistry changes in the surface and near-surface region. The technique provides quantitative elemental and chemical information with extremely high surface specificity and is ideal for comprehensively and quantitatively characterizing the elemental composition and chemical bonding states in graphene and carbon nanotubes.
Our webinar demonstrates how using both techniques in concert allows analysts to completely characterize carbon nanomaterials as illustrated by examples from graphene samples created by mechanical exfoliation, chemical reduction and CVD methods.
Areas of Interest:
X-ray photoelectron spectroscopy delivers chemical state information from the topmost few nanometers of the surface of a sample. Modern instruments, such as the Thermo Scientific™ K-Alpha™+ XPS system, can extend analyses into chemical images of the surface, revealing the extent of surface modification, or allowing defect detection. These strengths allow scientists and engineers to measure the effectiveness of coatings, understand plasma modification processes, and develop bio-compatibility coatings.
This webinar covers the basics of X-ray photoelectron spectroscopy, with a special emphasis on how it can be
used in the field of metal and oxide analysis.
This webinar is an introduction to Parallel Imaging and an overview of the benefits of using it to investigate surface chemistry. Learn to identify surface chemistry from small features using the combination of the XPS technique and Parallel Imaging hardware.
Access a targeted collection of application notes, case studies, videos, webinars and white papers covering a range of applications for Fourier Transform infrared spectroscopy, Near-infrared spectroscopy, Raman spectroscopy, Nuclear Magnetic Resonance, Ultraviolet-Visible (UV-Vis) spectrophotometry, X-Ray Fluorescence, and more.
Related spectroscopy, elemental, and isotope analysis learning topics
More Learning Centers ›