Search
Take a closer look at the features and specifications of Thermo Scientific Heratherm Heating and Drying ovens and Thermo Scientific Vacuutherm and Lindberg Blue/M Vacuum Ovens.
![]() |
![]() |
![]() |
![]() |
Heratherm Advanced Protocol Security Ovens Safety features offer protection of your most precious samples |
Heratherm Advanced Protocol Ovens Ultimate accuracy and increased heating |
Heratherm General Protocol Ovens For everyday heating and drying applications |
Precision Compact Ovens Compact ovens for drying and baking applications |
| Temperature range | |||
| 50 – 330°C | 50 – 330°C | 50 – 250°C | +5 – 210°C |
| Capacity | |||
| 61 – 170 L models | 61 – 731 L models | 65 – 744 L models | 48 L models |
| Heating options | |||
| Mechanical and gravity convection | |||
| Programmability | |||
|---|---|---|---|
10 automatically repeatable programs, each with up to 10 discrete steps. Access port for remote monitoring. |
10 automatically repeatable programs, each with up to 10 discrete steps | None | |
| Ordering information | |||
| All regions: models | Americas: 120V 60Hz models Europe, Africa, Asia: 230V 60Hz models Japan: 208-240V 60Hz, 208-240V 60Hz |
Americas: 120V 60Hz Europe, Africa, Asia: 230V 50/60Hz |
Americas only: 120V |
![]() |
![]() |
![]() |
Vacutherm Vacuum Ovens Achieve precise temperatures for simple or complex drying processes including heat sensitive materials |
Lindberg/Blue M Vaccum Ovens Designed for life science, industrial, and general lab applications requiring higher performance standards |
Thermo Scientific Vacuum Ovens For everyday use including drying, curing, vacuum embedding, and plating applications |
| Temperature range | ||
| Up to 400 °C | 6 – 260 °C | Ambient +10—220 °C |
| Capacity | ||
| 25 – 128 L | 18.6 – 42.5 L | 12.5 – 65 L options |
| Heating options | ||
| Jacket heating or shelf conduction | Jacket heating | Jacket heating |
| Safety features | ||
|---|---|---|
Overtemperature sensor protects oven and sample Double-pane safety glass doors Access port in rear wall for remote monitoring |
Independent overtemperature safety system factory set with reset pushbutton | Polycarbonate safety shield protects door glass |
| Ordering information | ||
| View models | View models | View models |
![]() |
![]() |
![]() |
Heratherm Heating and Drying Oven |
Vacutherm Vacuum Oven |
Lindberg Blue/M Vacuum Oven Accessories |
| Available accessories | ||
|
|
|
| All Heratherm Oven Shelf options | All Vacutherm Vacuum Accessories | All Linderberg/Blue M Vacuum Oven Accessories |