Search
Eureka Analysis Suite manages samples, ensures next-generation sequencing (NGS) index combination integrity, and enables genotyping data analysis with a single software package. Eureka Analysis Suite integrates sample management and tracking, single-nucleotide polymorphism (SNP) genotyping, and insertion/deletion (indel) detection in an easy-to-use graphical interface. The software automates the Best Practices Workflow to deliver accurate results in a single step for export to PLINK, VCF, or TXT formats.
Manage samples and run complete genotyping analysis of all your Eureka genotyping assays with a single software package.
Sample tracking
For a list of functions, please download the latest release notes.
Download the latest version of Eureka Analysis Suite, version 1.0 (64-bit), and install the software by following the installation instructions found in the Eureka Analysis Suite User Guide. This version is supported on Microsoft Windows™ 7 Professional (SP 1), Quad Core System 2.83 GHz with recommended 16 GB RAM.
For Research Use Only. Not for use in diagnostic procedures.