Search
An addition reaction occurs when two molecules are joined to create a more complex compound; all of the atoms in the original molecules are usually incorporated into the larger one. Electrophilic addition involves the attack of a primary substrate by an electrophile. The substrate generally possesses a carbon-carbon double or triple bond.
One of the first named reactions featuring electrophilic addition was the Prilezhaev reaction, named after the Russian chemist Nikolai Alexandrovich Prilezhaev, who in 1909, was the first to use peroxycarboxylic acids to oxidize isolated double bonds to the corresponding oxiranes. Because of its wide utility, epoxidation is one of the most frequently used reactions in organic chemistry. Epoxides such as epichlorohydrin are used in the manufacture of epoxy resins, a group of adhesives.
Learn more about the most popular named reactions based on feedback from our customers. Do you have a favorite reaction? Let us know!
Vote nowFor Research Use Only. Not for use in diagnostic or therapeutic procedures.