Search
AlleleTyper™ Software is a fast and easy tool that is used to determine the haplotypes of your samples. Once a translation table is set up for a study of interest, such as CYP2D6, AlleleTyper™ Software will analyze your TaqMan® Genotyper and CopyCaller® data in just a few quick steps. AlleleTyper™ Software interprets your real-time PCR analysis data and determines the star-allele results for your study based on your specifications.
AlleleTyper™ Software features:
AlleleTyper™ translator templates
Monoallelic tables and biallelic translator templates are provided for a few commonly researched genes including CYP2D6, CYP2C9, CYP2C19. These are provided as a starting point,and will likely need to be edited for specific studies. Refer to the AlleleTyper™ user guide for instructions on editing and using these types of files.
Download the translator templates >
Example Data Files
AlleleTyper™ interprets data that has been exported from the TaqMan® Genotyper™ and CopyCaller® Software. Examples of this data can be downloaded below. These data files can be used with the provided translator templates (see above section) to generate example results in AlleleTyper™ and helps to demonstrate the workflow. Refer to the AlleleTyper™ User Guide for assistance.
Download the example data files >
Note: AlleleTyper™ Software requires an internet connection and is only accessible through a compatible web browser. Google Chrome is recommended for optimal performance