Search
There are 100’s of different applications for measuring pH. When you consider the number of available instruments, electrodes, calibration standards, and individual techniques, it might be hard to find two users that measure pH the exact same way. Some users are able to achieve the best possible results, while many more users struggle to get accurate and repeatable pH measurements.
Our 7 tips for perfect pH will help to take the guesswork out of your pH measurements, helping you get the most out of your pH system whether you have a pocket tester or an advanced benchtop meter.
![]()
Cleaning a dirty, clogged or coated electrode can restore proper electrode performance and prolong the useful life of the electrode. Use the correct cleaning solution appropriate to the samples being measured.
![]()
A pH system is only as accurate as the calibration buffers that are used for standardization.
For best accuracy, use a minimum of two-point standardization; first with a buffer value close to the electrode systems zero potential (typically pH 7); and next with an acid or base buffer whose value brackets the expected pH value of the sample(s).
![]()
Use distilled water or electrode rinse solution to thoroughly rinse the electrode and never rub or blot the bulb to dry it. Gently shake off the excess water.
Electrode bulbs should always remain hydrated, and a dry electrode bulb will cause more problems than one or two drops of water in your sample.
![]()
If you use a refillable electrode be sure to check your fill level before storing and measurement.
Ensure that the filling hole is closed for storage, but open for calibration and measurement.
![]()
The temperature of your sample can have an effect on the pH. Ensure the temperature has stabilized on the probe before taking a reading and always record this along with the pH.
![]()
pH values of buffers at various temperatures | |||||||
|---|---|---|---|---|---|---|---|
| 0˚C | 1.67 | 4.00 | 6.98 | 47.11 | 9.46 | 10.32 | 12.79 |
| 5˚C | 1.67 | 4.00 | 6.95 | 7.08 | 9.40 | 10.25 | 12.73 |
| 10˚C | 1.67 | 4.00 | 6.92 | 7.06 | 9.33 | 10.18 | 12.67 |
| 20˚C | 1.67 | 4.00 | 6.87 | 7.01 | 9.23 | 10.06 | 12.52 |
| 25˚C | 1.68 | 4.01 | 6.86 | 7.00 | 9.18 | 10.01 | 12.46 |
| 30˚C | 1.68 | 4.02 | 6.85 | 6.98 | 9.14 | 9.97 | 12.36 |
| 40˚C | 1.69 | 4.04 | 6.84 | 6.97 | 9.07 | 9.89 | 12.17 |
| 50˚C | 1.71 | 4.06 | 6.83 | 6.97 | 9.01 | 9.83 | 11.96 |
| 60˚C | 1.72 | 4.09 | 6.84 | 6.97 | 8.96 | 9.79 | 11.73 |
| 70˚C | 1.74 | 4.13 | 6.85 | 6.99 | 8.92 | 9.78 | 11.47 |
| 80˚C | 1.77 | 4.16 | 6.86 | 7.03 | 8.89 | 9.78 | 11.19 |
| 90˚C | 1.79 | 4.21 | 6.88 | 7.08 | 8.85 | 9.80 | 10.89 |
The slope should be greater than 92% (53.5 mV/pH @ 20°C or 54.4 mV/pH @ 25°C). The offset (as measured in fresh pH 7 buffer) should be +/- 30 mV for new electrodes and +/- 59 mV for aged electrodes to enable auto-buffer recognition during calibration.
![]()
| Slope | at 20°C | at 25°C | Status | ||
|---|---|---|---|---|---|
| 1 pH unit | 3 pH unit | 1 pH unit | 3 pH unit | ||
| >102 | >59.3 | >178.0 | >60.3 | >181.0 | Poor |
| 102% | 59.3 | 178.0 | 60.3 | 181.0 | OK |
| 101% | 58.8 | 176.3 | 59.8 | 179.3 | Very Good |
| 100% | 58.2 | 174.5 | 59.2 | 177.5 | Ideal |
| 99% | 57.6 | 172.8 | 58.6 | 175.7 | Very Good |
| 98% | 57.0 | 171.0 | 58.0 | 173.9 | Very Good |
| 97% | 56.4 | 169.3 | 57.4 | 172.2 | Very Good |
| 96% | 55.8 | 167.5 | 56.8 | 170.4 | Very Good |
| 95% | 55.3 | 165.8 | 56.2 | 168.6 | Very Good |
| 94% | 57.0 | 171.0 | 58.0 | 173.9 | OK |
| 93% | 54.1 | 162.3 | 55.0 | 165.1 | OK |
| 92% | 53.5 | 160.5 | 54.4 | 163.3 | OK |
| <92 | <53.5 | <160.5 | <54.4 | <163.3 | Poor |
![]()
Cleaning a dirty, clogged or coated electrode can restore proper electrode performance and prolong the useful life of the electrode. Use the correct cleaning solution appropriate to the samples being measured.
![]()
A pH system is only as accurate as the calibration buffers that are used for standardization.
For best accuracy, use a minimum of two-point standardization; first with a buffer value close to the electrode systems zero potential (typically pH 7); and next with an acid or base buffer whose value brackets the expected pH value of the sample(s).
![]()
Use distilled water or electrode rinse solution to thoroughly rinse the electrode and never rub or blot the bulb to dry it. Gently shake off the excess water.
Electrode bulbs should always remain hydrated, and a dry electrode bulb will cause more problems than one or two drops of water in your sample.
![]()
If you use a refillable electrode be sure to check your fill level before storing and measurement.
Ensure that the filling hole is closed for storage, but open for calibration and measurement.
![]()
The temperature of your sample can have an effect on the pH. Ensure the temperature has stabilized on the probe before taking a reading and always record this along with the pH.
![]()
pH values of buffers at various temperatures | |||||||
|---|---|---|---|---|---|---|---|
| 0˚C | 1.67 | 4.00 | 6.98 | 47.11 | 9.46 | 10.32 | 12.79 |
| 5˚C | 1.67 | 4.00 | 6.95 | 7.08 | 9.40 | 10.25 | 12.73 |
| 10˚C | 1.67 | 4.00 | 6.92 | 7.06 | 9.33 | 10.18 | 12.67 |
| 20˚C | 1.67 | 4.00 | 6.87 | 7.01 | 9.23 | 10.06 | 12.52 |
| 25˚C | 1.68 | 4.01 | 6.86 | 7.00 | 9.18 | 10.01 | 12.46 |
| 30˚C | 1.68 | 4.02 | 6.85 | 6.98 | 9.14 | 9.97 | 12.36 |
| 40˚C | 1.69 | 4.04 | 6.84 | 6.97 | 9.07 | 9.89 | 12.17 |
| 50˚C | 1.71 | 4.06 | 6.83 | 6.97 | 9.01 | 9.83 | 11.96 |
| 60˚C | 1.72 | 4.09 | 6.84 | 6.97 | 8.96 | 9.79 | 11.73 |
| 70˚C | 1.74 | 4.13 | 6.85 | 6.99 | 8.92 | 9.78 | 11.47 |
| 80˚C | 1.77 | 4.16 | 6.86 | 7.03 | 8.89 | 9.78 | 11.19 |
| 90˚C | 1.79 | 4.21 | 6.88 | 7.08 | 8.85 | 9.80 | 10.89 |
The slope should be greater than 92% (53.5 mV/pH @ 20°C or 54.4 mV/pH @ 25°C). The offset (as measured in fresh pH 7 buffer) should be +/- 30 mV for new electrodes and +/- 59 mV for aged electrodes to enable auto-buffer recognition during calibration.
![]()
| Slope | at 20°C | at 25°C | Status | ||
|---|---|---|---|---|---|
| 1 pH unit | 3 pH unit | 1 pH unit | 3 pH unit | ||
| >102 | >59.3 | >178.0 | >60.3 | >181.0 | Poor |
| 102% | 59.3 | 178.0 | 60.3 | 181.0 | OK |
| 101% | 58.8 | 176.3 | 59.8 | 179.3 | Very Good |
| 100% | 58.2 | 174.5 | 59.2 | 177.5 | Ideal |
| 99% | 57.6 | 172.8 | 58.6 | 175.7 | Very Good |
| 98% | 57.0 | 171.0 | 58.0 | 173.9 | Very Good |
| 97% | 56.4 | 169.3 | 57.4 | 172.2 | Very Good |
| 96% | 55.8 | 167.5 | 56.8 | 170.4 | Very Good |
| 95% | 55.3 | 165.8 | 56.2 | 168.6 | Very Good |
| 94% | 57.0 | 171.0 | 58.0 | 173.9 | OK |
| 93% | 54.1 | 162.3 | 55.0 | 165.1 | OK |
| 92% | 53.5 | 160.5 | 54.4 | 163.3 | OK |
| <92 | <53.5 | <160.5 | <54.4 | <163.3 | Poor |
The ideal choice for dedicated pH testing in water, process, research, and QA/QC samples. The Thermo Scientific portfolio of pH Meters offer a range from simple & economical models to powerful & sophisticated pH meters, to help you analyze your samples with confidence.
Our portable pH meters are designed to offer pH testing in the field for applications including environmental, drinking water, and wastewater analysis.
The Thermo Scientific electrodes portfolio spans a vast range of industries and applications, and are categorised into digestible classifications to help you choose which electrode is best for your application.
The quality of your measurement results depends upon the quality of your standards, buffers, and reagents. We offer pH buffers and test kits for a variety of applications.