The use of a PCR Master Mix that does not contain ROX™ dye (or a similar passive reference) can cause:
- Trailing clusters
- Some or all data is undetermined (There is an "X" instead of a called allele in the AD plot.)
- Diffuse clusters
Solution:
- ROX™ dye is a passive reference dye that improves the precision of the results by compensating for small fluorescent fluctuations, such as bubbles and small well-to-well variations.
Note: The Sequence Detection Software on the 7900HT Fast Real-Time PCR System will not call the alleles when ROX™ dye (or another passive reference) is not present.
- Use Applied Biosystems TaqMan® Genotyping Master Mix or TaqMan® Universal PCR Master Mix, both of which contain ROXÔ passive reference.
- Confirm ROXÔ passive reference is specified in SDS software:
Specify ROX™ passive reference in SDS software