Search
Industrial and advanced materials characterization facilities frequently encounter unknown materials and diverse requests, necessitating comprehensive analytical capabilities to address varying material types and meet requirements for speed, productivity, and result reliability. Energy dispersive X-ray spectroscopy (EDS) offers an efficient solution for obtaining chemical information at the macro, micro, and nano scales with minimal preparation. EDS facilitates the identification and mapping of elemental composition within the material of interest, making it a valuable tool for these facilities.
The battery production cycle is a complex process that requires intermediate inspections to ensure production system quality. Checks include the inspection of raw materials and production of intermediate components.
Contamination poses a significant concern in battery manufacturing, potentially leading to reduced materials efficiency, accelerated cell degradation, and internal shorts. EDS serves as a crucial tool in battery research for detecting both intended materials and contaminants that can impact performance and safety.
Polymers are essential in our daily lives and in various industries, including household products, textiles, transportation, aviation, and aerospace. Many applications demand exceptional material properties, making it crucial to thoroughly study and optimize polymer microstructures for optimal performance.
Analytical methods like EDS are indispensable for characterizing and analyzing polymers. EDS offers valuable insights into the composition of industrial and newly developed materials, assists in identifying foreign elements to trace contamination in production, and aids in failure analysis to improve material performance).
Contemporary advanced metals are increasingly engineered at the nanoscale to improve durability, reliability, and cost-effectiveness. Even conventional methods now incorporate microscopic inspection to assess the elemental and structural composition of the resulting material. Precise control of inclusions and precipitates is essential for efficient metal production.
EDS plays a vital role in characterizing the chemical and structural aspects of these microscopic properties. Depending on the consistency and distribution of inclusions and precipitates, they can either strengthen the material or serve as contaminants, significantly affecting quality and lifespan.
Accurate analysis of texture and the associated distribution of minerals within the rock is vital to comprehensively describing the physical and chemical aspects of a rock system. EDS serves various purposes in geological research, including mineral identification, elemental analysis, petrography, geochemical mapping, and diagenetic studies. EDS offers high spatial resolution and non-destructive analysis, making it ideal for fragile or valuable geological samples. It is the preferred technique for identifying major and trace element compositions in minerals.
EDS enables elemental mapping, visually representing mineral phase distribution and providing insights into geological history and depositional environments. EDS also plays a critical role in studying diagenesis, including mineral alterations and new mineral formation, as well as petrography, helping infer rock formation conditions and subsequent processes.
Nanoparticles play a significant role in modern industries. They are used across various sectors to enhance mechanical, thermal, and optical material properties through incorporation into materials. They are utilized in medicine, energy, aerospace, cosmetics, textiles, and more, serving purposes like coatings, drug delivery, and material improvement.
Precisely quantifying nanoparticle composition, sizes, and shapes is an essential initial step in understanding their unique attributes. Energy-dispersive X-ray spectroscopy (EDS) in both SEMs and TEMs enables researchers to acquire nanometer- and sub-nanometer-scale images and chemical data, offering valuable insights into nanoparticle arrangement and potential functions.
The morphology, distribution, size, and chemical composition of nanoparticles significantly influence their catalytic efficiency. The combination of scanning transmission electron microscopy (S/TEM) and energy-dispersive X-ray spectroscopy (EDS) serves as a valuable research tool for directly observing and quantifying these factors. Furthermore, high-performance scanning electron microscopy (SEM) tools can capture high-quality images of beam-sensitive catalyst materials under low-beam-energy and low-beam-current conditions, all without causing sample damage.
For Research Use Only. Not for use in diagnostic procedures.