Select the ideal human leukocyte antigen (HLA) typing solution for your lab
Our broad portfolio of typing products offer low to high resolution detection to identify and match HLA. Assays include those that have revolutionized the workflow by putting the most efficient next-generation sequencing method into the hands of laboratory professionals.
One Lambda HybriType NGS assay
AllType NGS assays
LinkSēq PCR typing kits
Explore our HLA typing products
One Lambda HybriType NGS assay
A probe-based hybrid capture assay for eleven HLA and seven non classical loci with the option of ABO genotyping. Universal protocol for whole blood and buccal swabs with reduced hands- on and turnaround time.
AllType NGS assays
Multiplex PCR for genotyping for eleven HLA Class I and Class II loci with a single tube amplification for with short and long-range sequencing options. Streamlined workflow on multiple platforms with minimal hands-on time.
LABType assays
Combining Luminex xMAP technology with reverse sequence-specific oligonucleotide (rSSO) technology, high throughput DNA typing is achieved.
LinkSēq PCR typing kits
The LinkSēq line of real-time PCR products offers rapid genotyping of HLA and KIR with a simple workflow.
Micro SSP typing trays
Amplify genomic DNA using SSP to determine HLA Class I and Class II alleles.
SeCore kits
High-resolution typing of HLA alleles using SBT technique.
Genotyping assays
Cytokine, HNA, and KIR genotyping assays.
Serology reagents
Our tissue typing family offers a host of HLA testing reagents optimized for use with our HLA typing trays.
Lambda Monoclonal Trays (LMT)
Our exclusive LMT feature premixed monoclonal antibodies and rabbit complement for a fast and dependable method of determining Class I or Class II antigens.
Terasaki HLA tissue typing trays
Terasaki HLA Tissue Typing Trays contain well-characterized antisera that produce consistent and accurate HLA typing with complement-dependent cytotoxicity methods.
Product support and software files
⠀
Access One Lambda software, product documentation, and support files                                                   
Not all products are CE marked or have 510(k) clearance for sale in the U.S. Availability of products in each country depends on local regulatory marketing authorization status.