Search
From targeted biomarker assays to proteomics, metabolomics, and lipidomics, Thermo Scientific OptiSpray technology is designed to simplify LC-MS data acquisition and reduce common setup and troubleshooting challenges. OptiSpray technology is an intelligent, easy-to-use electrospray ionization (ESI) interface that combines automated spray optimization with cartridge capillary and nano LC columns, eliminating the need for fragile columns, emitters, and connections, simplifying setup and use, and improving quantitative accuracy and precision.
Plug-and-play setup and cartridge installation helps eliminate the challenge of manual setup.
“Our LC-MS systems are more approachable thanks to OptiSpray technology. Our researchers benefit from its intuitive design, enabling them to quickly produce robust proteomics results without extensive training or complex troubleshooting. This ion source is a critical innovation towards making proteomics more accessible for a wider range of laboratories.”
Ryan Lieb, Ph.D.
Director, Stanford University Mass Spectrometry
Automated emitter position optimization yields reproducible, high-quality results.
“In my view, OptiSpray technology is a game-changer. For decades, nano-LC-MS has been essential for ultra-sensitive protein analysis, yet its broad adoption in quantification has been limited by serious concerns on reproducibility and robustness. With OptiSpray technology, we’ve seen exceptional run-to-run reproducibility across thousands of biological samples, performance we’ve never observed on other platforms, which is critical for clinical and pharmaceutical research.”
Jun Qu, Ph.D.
Professor, University at Buffalo
Exchangeable emitters, integrated sheath gas, and an inactive spray position between injections enhance robustness, quantitative precision, and reproducibility.
“OptiSpray technology democratizes electrospray ionization by eliminating the skill gap in emitter positioning, enabling routine users to achieve performance comparable to experienced mass spectrometrists.”
Thomas Conrads, Ph.D.
Director of Women’s Health Research, Inova Women’s Service Line
Our OptiSpray family of products includes an ion source, cartridge columns, emitters, and accessories.
The Thermo Scientific OptiSpray Ion Source is an all-in-one ESI source for mass spectrometer calibration, compound optimization, and data acquisition at capillary and nano LC flow rates that combine automated setup and optimization with state-of-the-art performance.
Peace of mind through green and white light visualization of electrospray and sensitive measurement of spray current stability.
The OptiSpray ion source slides the emitter to an inactive spray position away from the MS inlet during column washing, keeping your mass spectrometer cleaner for longer.
Thermo Scientific OptiSpray Cartridge Columns provide robust and reproducible separations for your high-sensitivity LC-MS workflows. Cartridge columns are available in multiple phases and dimensions, including Thermo Scientific PepMap Neo and µPAC Neo C18 Columns.
Automatic fluidic, gas, and electrical connections upon cartridge installation, improving performance, reproducibility, and robustness.
Extended column lifetime from hundreds to thousands of samples with replaceable emitters tailored to both high-sensitivity nano-flow and highly robust capillary-flow.
Improved spray stability and quantitative precision for capillary-flow methods.
Memory stores cartridge properties, usage data, and previous source settings to get you up and running fast when switching cartridges.
The infusion cartridge provides simple mass spectrometer calibration and direct infusion while the source diagnostic cartridge supports intuitive diagnostics and troubleshooting.
OptiSpray technology consists of two parts—the OptiSpray ion source and OptiSpray cartridge columns. These two pieces combine to deliver a plug-and-play solution for high-sensitivity capillary and nano LC-MS workflows including proteomics, metabolomics, lipidomics as well as translational research and targeted assays.
OptiSpray technology offers several advantages compared with EASY-Spray sources including, but not limited to:
OptiSpray technology is advantageous for a wide range of reversed-phase LC-MS applications that require high-sensitivity including (but not limited to) all proteomics applications, metabolomics, lipidomics, and a range of other peptide and small molecule targeted applications.
OptiSpray technology is compatible with many Thermo Scientific mass spectrometers including:
Our general purpose product lines, including LC-MS products, are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks. They are designated for Research Use Only Applications.
For Research Use Only - Not For Use In Diagnostic Procedures.