LC-SPDP (succinimidyl 6-(3(2-pyridyldithio)propionamido)hexanoate)
Thermo Scientific™

LC-SPDP (succinimidyl 6-(3(2-pyridyldithio)propionamido)hexanoate)

Thermo Scientific Pierce LC-SPDP is a long-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavableRead more
Have Questions?
Catalog NumberQuantity
2165150 mg
Catalog number 21651
Price (USD)
594.00
Each
Quantity:
50 mg
Price (USD)
594.00
Each
Thermo Scientific Pierce LC-SPDP is a long-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls.

Features of LC-SPDP:

• Reactive groups: NHS ester and pyridyldithiol
• Reactive towards: amino and sulfhydryl groups
• Releases a detectable by-product when reacted with a free sulfhydryl group; by measuring the release of pyridine-2-thione at 343 nm, the reaction can be easily followed
• Disulfide bond in the spacer arm is readily cleaved by 10-50 mM DTT or TCEP at pH 8.5
• Spacer arm is also easily cleaved using reducing SDS-PAGE sample loading buffer
• Cleavable crosslinker allows separation of crosslinked products
• Water-insoluble (dissolve first in DMF or DMSO)
• SPDP crosslinker is membrane permeable, so crosslinking can be done inside the cells
• Compare to other varieties of SPDP-type reagents, including pegylated forms

Applications:
• Reversible crosslinking of proteins
• Adding sulfhydryl groups to proteins by attachment to lysine residues
• Protein crosslinking to create antibody-enzyme and other bioconjugates via a two-step reaction
• Attaching haptens to carrier proteins for antibody production
• Preparing immunotoxin conjugates

Specifications:
We manufacture LC-SPDP to the highest specification to produce the most specific bioconjugate, ensures the integrity of your data and to provide you with the highest degree of consistency. Each lot of LC-SPDP is tested to ensure it is >95% pure by quantitative NMR (the highest standard for crosslinker purity).

We offer three analogs of SPDP: the standard version (SPDP), a derivative with a longer spacer arm (LC-SPDP), and a sulfonated water-soluble variety (Sulfo-LC-SPDP). SPDP (N-Succinimidyl 3-(2-pyridyldithio)-propionate) and LC-SPDP (Succinimidyl 6-(3-[2-pyridyldithio]-propionamido)hexanoate) are heterobifunctional, thiol-cleavable and membrane permeable crosslinkers. They each contain an amine-reactive N-hydroxysuccinimide (NHS) ester that will react with lysine residues to form a stable amide bond. The other end of the spacer arm is terminated in the pyridyl disulfide group that will react with sulfhydryls to form a reversible disulfide bond.

Product References:
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
SPDP (succinimidyl 3-(2-pyridyldithio)propionate)
Pierce™ Premium Grade SPDP
Sulfo-LC-SPDP (sulfosuccinimidyl 6-(3'-(2-pyridyldithio)propionamido)hexanoate)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityYes
DescriptionLC-SPDP
FormPowder
Labeling MethodChemical Labeling
Molecular Weight (g/mol)425.52
PEGylatedNo
Product LinePierce
Quantity50 mg
Reactive MoietyNHS Ester, Pyridyldisulfide
Shipping ConditionAmbient
SolubilityDMF, DMSO
Spacer Arm Length15.7 Å
Water SolubleNo
Chemical ReactivityAmine-Sulfhydryl
CleavableBy Thiols
Crosslinker TypeHeterobifunctional
FormatStandard
Product TypeCrosslinker
SpacerMedium (10 to 30 Å)
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C protected from moisture.

Frequently asked questions (FAQs)

Can you provide the shelf-life for LC-SPDP (succinimidyl 6-(3(2-pyridyldithio)propionamido)hexanoate)?

LC-SPDP (succinimidyl 6-(3(2-pyridyldithio)propionamido)hexanoate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (4)

Citations & References
Abstract
Evidence for the assembly of a bacterial tripartite multidrug pump with a stoichiometry of 3:6:3.
Authors:Janganan TK, Bavro VN, Zhang L, Matak-Vinkovic D, Barrera NP, Venien-Bryan C, Robinson CV, Borges-Walmsley MI, Walmsley AR
Journal:J Biol Chem
PubMed ID:21610073
'The multiple transferable resistance (mTR) pump from Neisseria gonorrhoeae MtrCDE multidrug pump is assembled from the inner and outer membrane proteins MtrD and MtrE and the periplasmic membrane fusion protein MtrC. Previously we established that while there is a weak interaction of MtrD and MtrE, MtrC binds with relatively high ... More
Resurrection of the flagellar rotary motor near zero load.
Authors:Yuan J, Berg HC
Journal:Proc Natl Acad Sci U S A
PubMed ID:18202173
Flagellated bacteria, such as Escherichia coli, are propelled by helical flagellar filaments, each driven at its base by a reversible rotary motor, powered by a transmembrane proton flux. Torque is generated by the interaction of stator proteins, MotA and MotB, with a rotor protein FliG. The physiology of the motor ... More
A periplasmic coiled-coil interface underlying TolC recruitment and the assembly of bacterial drug efflux pumps.
Authors:Lobedanz S, Bokma E, Symmons MF, Koronakis E, Hughes C, Koronakis V
Journal:Proc Natl Acad Sci U S A
PubMed ID:17360572
Bacteria such as Escherichia coli and Pseudomonas aeruginosa expel antibiotics and other inhibitors via tripartite multidrug efflux pumps spanning the inner and outer membranes and the intervening periplasmic space. A key event in pump assembly is the recruitment of an outer membrane-anchored TolC exit duct by the adaptor protein of ... More
The assembled structure of a complete tripartite bacterial multidrug efflux pump.
Authors:Symmons MF, Bokma E, Koronakis E, Hughes C, Koronakis V
Journal:Proc Natl Acad Sci U S A
PubMed ID:19342493
Bacteria like Escherichia coli and Pseudomonas aeruginosa expel drugs via tripartite multidrug efflux pumps spanning both inner and outer membranes and the intervening periplasm. In these pumps a periplasmic adaptor protein connects a substrate-binding inner membrane transporter to an outer membrane-anchored TolC-type exit duct. High-resolution structures of all 3 components ... More