Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD206 (MMR) Antibodies

          Invitrogen

          CD206 (MMR) Monoclonal Antibody (MR6F3), PE-Cyanine7, eBioscience™

          72 Published Figures
          62 References
          View all (84) CD206 (MMR) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD206 (MMR) Monoclonal Antibody (MR6F3), PE-Cyanine7, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (72)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 73

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD206 (MMR) Antibody (25-2061-80) in Flow

          Mouse resident peritoneal exudate cells were surface stained with Anti-Mouse F4/80 Antigen eFluor® 450 (Product # 48-4801-82) followed by fixation and permeabilization with the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00). Cells were then intracellularly stained with 0.125 µg of Rat IgG2b K Isotype Control PE-Cyanine7 (Product # 25-4031-82) (left) or 0.125 µg of Anti-Mouse CD206 (MMR) PE-Cyanine7 (right). Total cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)
          CD206 (MMR) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          25-2061-80

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.25 µg/test
          View 62 publications 62 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Mouse

          Host/Isotype

          Rat / IgG2b, kappa

          Recommended Isotype Control

          Rat IgG2b kappa Isotype Control (eB149/10H5), PE-Cyanine7, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          MR6F3

          Conjugate

          PE-Cyanine7 PE-Cyanine7 PE-Cyanine7
          • Alexa Fluor 488 
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Brilliant UV 563
          • eFluor 450
          • PE 
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Request custom conjugation

          Excitation/Emission Max

          569/780 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2637423

          Product Specific Information

          Description: This MR6F3 antibody recognizes mouse CD206 also known as Macrophage Mannose Receptor (MMR) or Mannose Receptor C, Type 1 (MRC1). CD206 is a 175-kDa, type 1 integral membrane glycoprotein receptor that is present in macrophages, some dendritic cells, as well as liver and lymphoid endothelial cells. CD206 belongs to the C-type lectin family. Extracellular regions of CD206 include an N-terminal cysteine-rich (CR) domain that binds sulfated glycoproteins, a fibronectin II (FNII) domain that binds collagens, and eight carbohydrate recognition domains (CRDs) that bind sugars like mannose and fucose with high affinity. CD206 mediates phagocytic and endocytic uptake of fungal, bacterial, protozoan and viral antigens, and plays an important role in immune defense and immune regulation. A soluble form of CD206 is generated by cleavage of the full-length protein, and it can be detected in in vitro macrophage cell culture supernatants and in mouse serum. CD206 is considered to be one of the markers of M2 macrophages. Factors inducing its expression include: IL-4, IL-13, M-CSF, IL-6, IL-10, and glucocorticoids, while TNF-alpha, IFN-gamma, TGF-beta, and LPS have been reported to down-regulate expression of CD206.

          It has been demonstrated that only a small fraction of CD206 is present at the cell surface, therefore, intracellular staining is recommended.

          Applications Reported: This MR6F3 antibody has been reported for use in flow cytometric analysis, and intracellular staining followed by flow cytometric analysis.

          Applications Tested: This MR6F3 antibody has been tested by intracellular staining and flow cytometric analysis of mouse resident peritoneal exudate cells using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00 and protocol. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.

          Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

          Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CD206 (MSR, Mannose receptor, MRC1) is a 175 kDa transmembrane protein belonging to the group of pattern recognition receptors. CD206 is predominantly expressed in tissue macrophages, dendritic cells, a subpopulation of endothelial cells and sperm cells. CD206 is thought to play a role in the innate and adaptive immune response. CD206 is also expressed on microglia and mato cells of the brain but not astrocytes or neurons. CD206 also mediate the recognition and uptake of a variety of macromolecules, including modified lipoproteins, advanced glycation end (AGEs) products and amyloid b-protein (Abeta). While the normal role of CD206 is associated with cell adhesion and host defense mechanisms, it also has been implicated in the development of atherosclerosis and Amyloid beta deposition in Alzheimer's disease (AD). CD206's gene encodes the class A macrophage scavenger receptors, which include three different types (1, 2, 3) generated by alternative splicing. The isoforms type 1 and type 2 are functional receptors and are able to mediate the endocytosis of modified low density lipoproteins (LDLs). The isoform type 3 does not internalize modified LDL (acetyl-LDL) despite having the domain shown to mediate this function in the types 1 and 2 isoforms. CD206 has an altered intracellular processing and is trapped within the endoplasmic reticulum, making it unable to perform endocytosis. The isoform type 3 can inhibit the function of isoforms type 1 and type 2 when co-expressed, indicating a dominant negative effect and suggesting a mechanism for regulation of scavenger receptor activity in macrophages. Other diseases associated with CD206 dysfunction include leprosy and Gaucher's Disease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: MMR; unnamed protein product

          View more View less

          Gene Aliases: CD206; MR

          View more View less

          UniProt ID: (Mouse) Q61830

          View more View less

          Entrez Gene ID: (Mouse) 17533

          View more View less

          Function(s)
          transmembrane signaling receptor activity protein binding mannose binding cargo receptor activity
          Process(es)
          receptor-mediated endocytosis cellular response to lipopolysaccharide cellular response to interferon-gamma cellular response to interleukin-4
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-7d6nq:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline