Pierce™ Anti-HA Agarose
Thermo Scientific™
Pierce™ Anti-HA Agarose
Thermo Scientific Pierce Anti-HA Agarose is an immunopurification and immunoprecipitation resin for the enrichment of HA-tagged proteins expressed in humanRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
261825 mL
261811 mL
Catalog number 26182
Price (USD)
2,247.65
Online exclusive
2,525.00
Save 277.35
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
2,247.65
Online exclusive
2,525.00
Save 277.35
Each
Thermo Scientific Pierce Anti-HA Agarose is an immunopurification and immunoprecipitation resin for the enrichment of HA-tagged proteins expressed in human in vitro protein expression systems, bacteria, yeast, and mammalian cells. The anti-HA antibody coupled to the resin is a high-affinity mouse IgG1 monoclonal antibody that recognizes the HA-epitope tag (YPYDVPDYA) derived from the human influenza hemagglutinin (HA) protein.
Features of Pierce Anti-HA Agarose
  • Specific—highly specific anti-HA monoclonal antibody enables high yield and high purity
  • Scalable—available in 1 and 5 mL settled resin package sizes to allow for smaller scale immunoprecipitations or larger scale purifications
  • Versatile—can be used in spin or gravity-based purification and enrichment methods
Pierce Anti-HA Agarose consists of highly specific monoclonal anti-HA antibody that is covalently immobilized on a crosslinked 4% beaded agarose support. The product is supplied as a 50% slurry in 0.1 M phosphate, 0.15 M NaCl, pH 7.2 with 0.05% sodium azide. The anti-HA agarose has 3.5 mg mouse anti-HA IgG1 monoclonal antibody conjugated per mL of settled agarose resin.
HA-tagged proteins are easily eluted from the resin using 0.1 M glycine (pH 2.0–2.8), 3 M NaSCN, or 50 mM NaOH, depending on the downstream application of the purified protein. For gentle elution, HA Synthetic Peptide is available for use in neutralization and to competitively elute HA-tagged fusion proteins from immobilized anti-HA affinity resin.
Related products
Pierce HA-Tag IP/Co-IP Kit
Pierce HA-Tag Magnetic IP/Co-IP Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConjugateHA-epitope tag (YPYDVPDYA)
Purification TargetAnti-HA Antibody
Quantity5 mL
Column TypeAgarose Resin, Affinity
FormatBottle
Product TypeAgarose
Unit SizeEach
Contents & Storage
5 mL of settle resin, provided as 10 mL of 50% slurry. Store at 2–8°C.